Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02596851_gH
          See other USP41P GE Assays ›
          Gene Symbol:
          USP41P ETFDH
          Gene Name:
          ubiquitin specific peptidase 41, pseudogene
          electron transfer flavoprotein dehydrogenase
          Gene Aliases:
          USP18B, USP41
          ETFQO, MADD
          Chromosome Location:
          Chr.22: 20363616 - 20377260 on Build GRCh38
          Chromosome Location:
          Chr.4: 158672296 - 158709623 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs02596851_gH
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: USP41P
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AJ583822.1 - 4 - 5 388 115
          AJ586979.1 - 3 - 4 325 115
          AK303463.1 - 2 - 3 234 115
          Gene Symbol: ETFDH
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA KY419095.1 - 2 - 3 234 115

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32105837 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11433324 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32105874 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32105875 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11433325 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32105871 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17258871 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 8967 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 21071 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P21283117 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P22618415 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P54553 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P21283064 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P03602167 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P21283136 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P21283064 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03602205 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P21283117 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03602168 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P21283104 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P21283119 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03602167 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P21283136 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P54553 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 142954 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 66758 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 167379 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 63398 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 29365 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 87041 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 7083 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P22618415 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P21283064 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P13957559 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P21283117 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03602167 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03602205 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03602168 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P21283136 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03602213 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P22620520 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03602219 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P13962822 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P22618589 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03602172 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P21283044 - -
          Agilent Human 1B Microarray G4111A A_32_P54553 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array XM_036729_at - -
          Affymetrix Human Gene 1.0 ST Array XM_001718645_at - -
          Affymetrix Human Gene 1.0 ST Array 8074608 - -
          Affymetrix Human Gene 1.0 ST Array 8074613 - -
          Affymetrix Human Gene 1.0 ST Array 8074612 - -
          Affymetrix Human Gene 1.0 ST Array 8074607 - -
          Affymetrix Human Gene 1.0 ST Array 8074609 - -
          Affymetrix Human Gene 1.0 ST Array 8074614 - -
          Affymetrix Human Gene 1.0 ST Array 8074611 - -
          Affymetrix Human Gene 1.0 ST Array 8074610 - -
          Affymetrix Human Gene 1.1 ST Array 8074606 - -
          Affymetrix Human Genome U219 Array 11761542_PM_at - -
          Affymetrix Human Genome U219 Array 11761542_at - -
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000292729.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000292729.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000292729.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000292729.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000292729.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000292729.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000292729.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000292729.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000454608.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000454608.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000454608.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000454608.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000454608.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000454608.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000454608.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000454608.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000454608.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000454608.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000470202.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000470202.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000486536.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000486536.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000486536.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000486536.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000486536.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000486536.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000486536.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008036.hg.1 ENST00000486536.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000292729.12 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000292729.12 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000292729.12 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000292729.12 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000292729.12 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000292729.12 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000292729.12 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000292729.12 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000454608.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000454608.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000454608.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000454608.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000454608.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000454608.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000454608.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000454608.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000454608.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000454608.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000470202.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000470202.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000486536.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000486536.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000486536.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000486536.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000486536.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000486536.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000486536.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200008036.hg.1 ENST00000486536.6 Ensembl
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1690365 AJ583822.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1690365 AJ586979.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1778487 - -
          HumanHT-12 v4 Expression BeadChip ILMN_1801931 - -
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1690365 AJ583822.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1690365 AJ586979.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1801931 - -
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1723635 - -
          HumanHT-12 V4.0 expression beadchip ILMN_1801931 - -
          HumanHT-12 V4.0 expression beadchip ILMN_1690365 AJ583822.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1690365 AJ586979.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1801931 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1723635 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1690365 AJ583822.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1690365 AJ586979.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip ILMN_37497 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_37497 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1801931 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1690365 AJ583822.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1690365 AJ586979.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1778487 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1690365 AJ583822.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1690365 AJ586979.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1801931 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1778487 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1690365 AJ583822.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1690365 AJ586979.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1801931 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline