| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922860 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922869 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922832 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922870 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922844 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922829 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922854 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04727516 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922886 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04727467 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922831 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922871 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922816 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922876 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04727471 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922853 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922864 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922843 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922872 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922834 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922893 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922847 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922830 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04727532 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922839 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922883 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922852 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04727529 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922824 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922868 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922846 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922842 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922881 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922895 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15815189 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922873 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922862 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922825 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922835 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922887 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922815 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922828 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04727472 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922892 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04727474 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922867 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922850 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04727493 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922889 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922849 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922848 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922851 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922894 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15815186 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922891 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922874 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04727477 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922845 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04727478 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922861 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922856 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04727476 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922865 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922837 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24922888 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
114296 |
AF126426.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
114296 |
AY358331.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
114296 |
BC050716.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
114296 |
CR602526.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
114296 |
EF043567.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
114296 |
NM_001048209.1 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
114296 |
NM_016522.2 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CGH Microarray 8x60K |
27052 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
367847 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3354201 |
- |
- |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_HS_44k_1230 |
- |
- |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_HS_44k_1231 |
- |
- |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_24_P931533 |
- |
- |
| Agilent Whole Human Genome Oligo Microarray 4x44K |
11891 |
- |
- |
| Agilent Human Genome CGH Microarray 44B |
A_14_P123340 |
- |
- |
| Agilent Human Genome CGH Microarray 44A |
A_14_P123340 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
37231 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P01477824 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_18_P15643419 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3354201 |
- |
- |
| SurePrint G3 Human GE Microarray G4851A |
A_33_P3354201 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04727532 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922874 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922876 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04727515 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922868 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922869 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922880 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922867 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922888 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922895 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922894 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922886 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04727516 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922884 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922889 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922883 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922893 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922887 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922881 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922872 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922864 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922891 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04727529 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922891 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04727516 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922880 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922867 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922889 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922893 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922895 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922881 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922868 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04727529 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04727515 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922883 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922887 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922869 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922872 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922876 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922888 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922886 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922894 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922864 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922884 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04727532 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24922874 |
- |
- |
| Whole Human Genome Oligo Microarray G4112A |
A_24_P931533 |
- |
- |
| Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P123340 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P123340 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P01477824 |
- |
- |
| Agilent Human Genome CGH Microarray 4x44K |
A_14_P123340 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K G4112F |
A_24_P931533 |
- |
- |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
29115 |
- |
- |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
94956 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P17542329 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P01477824 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P123340 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727527 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727500 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727520 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727498 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727512 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727567 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727568 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727539 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727477 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727515 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727541 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727531 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727548 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727513 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727571 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727574 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727506 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727525 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727483 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727543 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727533 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727486 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727536 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727502 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727553 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727576 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727546 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15815189 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727560 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727563 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727484 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727583 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727470 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15815187 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727475 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727510 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727508 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727489 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727495 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727479 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15815193 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727581 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727517 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727565 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727504 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727473 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727585 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727530 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727467 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04727558 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P123340 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P01477824 |
- |
- |