Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02597401_m1
          See other LRRC37A5P GE Assays ›
          Gene Symbol:
          LRRC37A5P
          Gene Name:
          leucine rich repeat containing 37 member A5, pseudogene
          Gene Aliases:
          C9orf29
          Chromosome Location:
          Chr.9: 111602831 - 111613553 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02597401_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LRRC37A5P
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_034087.1 2 - 3 809 112
          GenBank mRNA BC031236.1 - 2 - 3 811 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900011204.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09001485.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06900087 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06900066 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16253945 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16253944 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06900065 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06900071 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27243588 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06900077 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27243599 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06900083 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06900075 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06900063 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27243578 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06900089 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06900094 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06900069 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06900062 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06900093 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27243573 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06900070 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27243570 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27243591 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06900060 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27243596 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16253943 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27243590 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27243584 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06900081 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06900084 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16253941 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06900072 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3399438 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 5827 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3399438 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3412013 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18738954 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02158746 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18738941 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P32509396 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02158749 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18738953 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8163345 - -
          Affymetrix Human Gene 1.0 ST Array 8163342 - -
          Affymetrix Human Gene 1.0 ST Array 8163344 - -
          Affymetrix Human Gene 1.0 ST Array 8163343 - -
          Affymetrix Human Gene 1.0 ST Array 8163347 - -
          Affymetrix Human Gene 1.0 ST Array 8163346 - -
          Affymetrix Human Gene 1.1 ST Array 8163341 - -
          Affymetrix HT HG-U133+ PM Array Plate 1562803_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1569839_PM_s_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR09018429.hg.1 ENST00000374302.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09018429.hg.1 ENST00000374306.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09018429.hg.1 ENST00000536054.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09018429.hg.1 NR_034087.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09018429.hg.1 TC09001485.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09018424.hg.1 ENST00000374306.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09018424.hg.1 ENST00000536054.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09018424.hg.1 NR_034087.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09018424.hg.1 TC09001485.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09018428.hg.1 ENST00000374302.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09018428.hg.1 ENST00000374304.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09018428.hg.1 ENST00000374306.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09018428.hg.1 ENST00000536054.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09018428.hg.1 NR_034087.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09018428.hg.1 TC09001485.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0900011204.hg.1 ENST00000374304.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011204.hg.1 ENST00000374304.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011204.hg.1 ENST00000374304.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011204.hg.1 ENST00000536054.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011204.hg.1 ENST00000536054.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011204.hg.1 ENST00000536054.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011204.hg.1 ENST00000536054.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011204.hg.1 ENST00000580181.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011204.hg.1 ENST00000580181.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011204.hg.1 NR_034087.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011204.hg.1 NR_034087.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011204.hg.1 NR_034087.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011204.hg.1 NR_034087.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900176286.hg.1 ENST00000536054.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900176286.hg.1 NR_034087.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900176286.hg.1 ENST00000536054.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900176286.hg.1 NR_034087.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900176286.hg.1 TC0900011204.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900176291.hg.1 ENST00000536054.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900176291.hg.1 NR_034087.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900176291.hg.1 ENST00000536054.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900176291.hg.1 NR_034087.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900176291.hg.1 TC0900011204.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0900011204.hg.1 ENST00000374304.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011204.hg.1 ENST00000374304.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011204.hg.1 ENST00000374304.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011204.hg.1 ENST00000536054.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900011204.hg.1 ENST00000536054.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900011204.hg.1 ENST00000536054.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900011204.hg.1 ENST00000536054.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900011204.hg.1 ENST00000580181.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011204.hg.1 ENST00000580181.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011204.hg.1 NR_034087.1 Refseq
          Affymetrix Human Clariom D Assay TC0900011204.hg.1 NR_034087.1 Refseq
          Affymetrix Human Clariom D Assay TC0900011204.hg.1 NR_034087.1 Refseq
          Affymetrix Human Clariom D Assay TC0900011204.hg.1 NR_034087.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900176289.hg.1 ENST00000374304.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900176289.hg.1 ENST00000536054.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900176289.hg.1 ENST00000580181.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900176289.hg.1 NR_034087.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900176289.hg.1 ENST00000374304.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900176289.hg.1 ENST00000536054.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900176289.hg.1 ENST00000580181.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900176289.hg.1 NR_034087.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900176289.hg.1 TC0900011204.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900176292.hg.1 ENST00000536054.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900176292.hg.1 NR_034087.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900176292.hg.1 ENST00000536054.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900176292.hg.1 NR_034087.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900176292.hg.1 TC0900011204.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 1569839_s_at - -
          Human Genome U133 Plus 2.0 Array 1562803_at - -
          Human Genome U133 Plus 2.0 Array 1569839_PM_s_at - -
          Human Genome U133 Plus 2.0 Array 1562803_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569839_s_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562803_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569839_s_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569839_s_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562803_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569839_s_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562803_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562803_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569839_s_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569839_s_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569839_s_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562803_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562803_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569839_s_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562803_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562803_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569839_s_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562803_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562803_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562803_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569839_s_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569839_s_at6 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A037903 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A037903 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1755698 NR_034087.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1755698 BC031236.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1755698 NR_034087.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1755698 BC031236.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1755698 NR_034087.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1755698 BC031236.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1755698 NR_034087.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1755698 BC031236.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1755698 NR_034087.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1755698 BC031236.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline