Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02597671_s1
          See other OR1N1 GE Assays ›
          Gene Symbol:
          OR1N1
          Gene Name:
          olfactory receptor family 1 subfamily N member 1
          Gene Aliases:
          OR1-26, OR1N3, OR9-22
          Chromosome Location:
          Chr.9: 122526358 - 122527293 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02597671_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: OR1N1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012363.1 NP_036495.1 1 458 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900011441.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09001559.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene OR1N1

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          G-protein coupled receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06947646 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06947673 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06947657 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27292819 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06947692 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27292799 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27292804 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06947650 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06947653 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06947684 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27292790 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06947669 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27292820 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06947663 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16260857 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27292832 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06947659 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06947678 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06947666 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06947648 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06947682 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06947681 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27292824 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06947661 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06947670 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06947688 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06947675 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06947662 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06947651 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27292833 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27292806 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06947686 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27292816 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06947642 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27292835 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06947687 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27292801 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27292821 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06947667 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27292795 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06947647 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27292789 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3223739 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_2550 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_2551 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 16332 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P02174668 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3223739 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3223739 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947681 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947662 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947682 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947686 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947669 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947663 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27292816 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947684 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27292833 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27292820 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947678 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947666 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947673 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947687 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27292832 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27292801 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27292819 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27292821 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947679 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27292824 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947659 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27292799 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947670 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27292806 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947661 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27292804 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947692 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947673 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27292801 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27292819 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947692 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27292832 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27292799 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947659 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947669 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947682 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947666 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947684 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27292806 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27292820 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27292816 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27292833 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947686 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27292821 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27292804 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27292824 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947662 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947679 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947678 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947670 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947663 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947687 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947661 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06947681 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P06947689 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P06947658 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P06947655 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P06947684 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P06947682 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P06947686 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P06947673 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P06947662 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P06947675 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P06947670 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P06947679 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P06947659 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P06947666 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P06947680 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P06947691 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P06947677 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947609 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947599 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947629 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947641 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947624 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947600 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947686 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947610 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947658 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947632 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947680 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947615 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947619 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947647 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947691 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947673 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947625 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947655 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947662 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947627 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16260853 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947612 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947666 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16260856 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947637 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947675 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947650 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947614 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947645 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947692 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947689 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947617 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947684 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947679 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947670 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947659 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947635 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947677 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947622 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06947682 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_012363_at - -
          Affymetrix Human Gene 1.0 ST Array 8163959 - -
          Affymetrix Human Gene 1.1 ST Array 8163958 - -
          Affymetrix Human Genome U219 Array 11742606_at - -
          Affymetrix Human Genome U219 Array 11742606_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0900011441.hg.1 ENST00000304880.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011441.hg.1 ENST00000619659.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011441.hg.1 NM_012363.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09019580.hg.1 TC09001559.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09019580.hg.1 ENST00000304880.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09019580.hg.1 NM_012363.1 Refseq
          Affymetrix Human Clariom D Assay TC0900011441.hg.1 ENST00000304880.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900011441.hg.1 ENST00000619659.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011441.hg.1 NM_012363.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900177998.hg.1 ENST00000304880.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177998.hg.1 ENST00000619659.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177998.hg.1 NM_012363.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900177998.hg.1 ENST00000304880.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177998.hg.1 ENST00000619659.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177998.hg.1 NM_012363.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900177998.hg.1 TC0900011441.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A038004 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A038004 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1739138 NM_012363.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1739138 NM_012363.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1739138 NM_012363.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1739138 NM_012363.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1739138 NM_012363.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_15786-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1739138 NM_012363.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1739138 NM_012363.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 830152 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_15786 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1739138 NM_012363.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1739138 NM_012363.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1739138 NM_012363.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1739138 NM_012363.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1739138 NM_012363.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline