Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02597779_s1
          See other HCAR1 GE Assays ›
          Gene Symbol:
          HCAR1
          Gene Name:
          hydroxycarboxylic acid receptor 1
          Gene Aliases:
          FKSG80, GPR104, GPR81, HCA1, LACR1, TA-GPCR, TAGPCR
          Chromosome Location:
          Chr.12: 122727606 - 122730582 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02597779_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HCAR1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_032554.3 NP_115943.1 1 2785 100
          GenBank mRNA AF385431.1 - 1 2785 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as HTA transcript cluster(s) TC12002077.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HCAR1

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of defense response to virus by host
          G-protein coupled receptor signaling pathway
          mitophagy in response to mitochondrial depolarization
          xenophagy
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08659989 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29096617 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29096622 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29096607 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08659972 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29096627 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08659986 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08660016 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08659971 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08660015 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08659987 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29096585 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29096597 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29096608 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29096616 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08660010 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08659974 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08659991 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08660004 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08659997 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08659975 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08659979 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08659995 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29096587 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08660012 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08659983 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08659980 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08659992 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08659985 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29096592 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08659982 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08659993 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08660009 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29096606 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29096609 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29096611 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08659988 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29096584 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08659981 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 54362 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 13966 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 451528 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 253178 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3380472 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_2931 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_2932 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 43132 - -
          Agilent Human Genome CGH Microarray 44B A_14_P121045 - -
          Agilent Human Genome CGH Microarray 44A A_14_P121045 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 61846 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P02717377 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3380472 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3235776 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3380472 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08660008 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29096622 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08659995 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08659993 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08659997 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08659980 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08659985 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29096617 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29096587 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29096610 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08659988 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29096606 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08660010 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08659983 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08659982 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29096611 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08659989 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08659986 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08660004 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29096608 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29096616 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08659992 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08660003 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08660008 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08660004 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08660010 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08659980 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08659992 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29096587 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08660003 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29096616 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29096622 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08659989 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29096606 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29096611 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08659985 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08659995 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08659982 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08659988 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08659983 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08659997 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08659986 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29096608 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08659993 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29096610 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29096617 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P121045 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08660005 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08659986 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08659995 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08659978 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08659997 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08660007 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08660003 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08660008 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08659985 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08659980 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08659982 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08660002 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08659993 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08659987 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08659989 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08660010 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P121045 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P121045 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P394246 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 139337 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 63094 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P121045 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08660012 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16598851 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08660016 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08659982 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08659985 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08659978 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08660002 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08659954 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08659997 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08660015 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16598853 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08660005 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08660007 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08659987 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08659980 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08659953 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08659986 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08659995 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08659976 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08660010 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08659989 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08659993 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08660008 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08659951 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08659974 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08659956 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16598856 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P121045 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_032554_at - -
          Affymetrix Human Gene 1.0 ST Array 7967327 - -
          Affymetrix Human Gene 1.0 ST Array 7967326 - -
          Affymetrix Human Gene 1.0 ST Array 7967328 - -
          Affymetrix Human Gene 1.0 ST Array 7967330 - -
          Affymetrix Human Gene 1.0 ST Array 7967329 - -
          Affymetrix Human Gene 1.1 ST Array 7967325 - -
          Affymetrix HT HG-U133+ PM Array Plate 224131_PM_at - -
          Affymetrix Human Genome U219 Array 11731079_s_at - -
          Affymetrix Human Genome U219 Array 11731080_PM_at - -
          Affymetrix Human Genome U219 Array 11750278_PM_s_at - -
          Affymetrix Human Genome U219 Array 11750278_s_at - -
          Affymetrix Human Genome U219 Array 11731080_at - -
          Affymetrix Human Genome U219 Array 11731079_PM_s_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR12027372.hg.1 ENST00000432564.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12027372.hg.1 NM_032554.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12027372.hg.1 TC12002077.hg.1 HTA TC
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_032554_at - -
          Human Genome U95 Set 41934_at - -
          Human Genome U95 Set 41934_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A048126 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A048126 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1811272 AF385431.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1811272 AF385432.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2161848 NM_032554.3 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1811272 AF385431.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1811272 AF385432.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1811272 AF385431.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1811272 AF385432.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2161848 NM_032554.3 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2161848 NM_032554.3 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2161848 NM_032554.3 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_21314751-S AF385431.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_21314751-S AF385432.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_139290-S - -
          Illumina human-6 v1.0 expression beadchip 0005050575 - -
          Illumina humanRef-8 v1.0 expression beadchip 0005050575 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1811272 AF385431.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1811272 AF385432.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1811272 AF385431.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1811272 AF385432.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 1070112 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_139290 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2161848 NM_032554.3 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1811272 AF385431.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1811272 AF385432.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2161848 NM_032554.3 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2161848 NM_032554.3 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1811272 AF385431.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1811272 AF385432.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1811272 AF385431.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1811272 AF385432.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2161848 NM_032554.3 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline