Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02599057_s1
          See other SPANXN3 GE Assays ›
          Gene Symbol:
          SPANXN3
          Gene Name:
          SPANX family member N3
          Gene Aliases:
          CT11.8, SPANX-N3
          Chromosome Location:
          Chr.X: 143508735 - 143517475 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02599057_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SPANXN3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009609.3 NP_001009609.2 2 252 139
          XM_017029264.1 XP_016884753.1 2 372 139
          GenBank mRNA BC062750.1 - 256 6618011 139
          DQ336128.1 - 2 168 139
          DQ336129.1 - 2 168 139

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0X00011010.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC0X001445.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SPANXN3

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P12000187 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17406561 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P12000176 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32734598 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32734577 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P12000178 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32734597 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32734580 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32734595 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P12000177 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32734565 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32734605 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17406576 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32734599 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32734602 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P12000183 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P12000185 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32734574 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P12000186 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 156436 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 964441 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 572137 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 208070 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 386689 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P91830 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P91830 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 38962 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 48907 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P60394355 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P03784643 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P91830 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P91830 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32734599 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P12000187 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32734595 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32734604 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32734597 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P12000189 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17406576 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32734605 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P12000186 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32734602 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32734598 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17406578 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P12000187 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32734598 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17406578 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32734599 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32734604 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32734602 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32734605 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32734595 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P12000186 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17406576 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P12000189 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32734597 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P91830 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P03784643 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P12000184 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P12000188 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P12000186 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P12000191 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P12000189 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P12000182 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03784643 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P91830 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 149194 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 10828 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03784643 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P41826740 - -
          Agilent Human 1B Microarray G4111A A_32_P899557 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P12000188 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P12000186 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P12000178 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P12000182 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17406576 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P12000179 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P12000177 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P12000175 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P12000184 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P12000191 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P12000174 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P12000181 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03784643 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001009609_at - -
          Affymetrix Human Gene 1.0 ST Array 8175573 - -
          Affymetrix Human Gene 1.1 ST Array 8175572 - -
          Affymetrix Human Genome U219 Array 11742165_at - -
          Affymetrix Human Genome U219 Array 11742165_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0X00011010.hg.1 ENST00000370503.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011010.hg.1 ENST00000370503.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011010.hg.1 ENST00000370503.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011010.hg.1 ENST00000370503.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011010.hg.1 NM_001009609.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011010.hg.1 NM_001009609.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011010.hg.1 NM_001009609.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011010.hg.1 NM_001009609.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X019083.hg.1 TC0X001445.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR0X019083.hg.1 ENST00000370503.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X019083.hg.1 NM_001009609.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X019084.hg.1 TC0X001445.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR0X019084.hg.1 ENST00000370503.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X019084.hg.1 NM_001009609.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008623.hg.1 ENST00000431432.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008623.hg.1 ENST00000431432.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008623.hg.1 ENST00000431432.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008623.hg.1 ENST00000431432.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008623.hg.1 ENST00000431432.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008623.hg.1 ENST00000431432.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00174745.hg.1 ENST00000370503.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00174745.hg.1 NM_001009609.3 Refseq
          Affymetrix Human Clariom D Assay PSR0X00174745.hg.1 ENST00000370503.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00174745.hg.1 NM_001009609.3 Refseq
          Affymetrix Human Clariom D Assay PSR0X00174745.hg.1 TC0X00011010.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0X00008623.hg.1 ENST00000431432.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008623.hg.1 ENST00000431432.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008623.hg.1 ENST00000431432.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008623.hg.1 ENST00000431432.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008623.hg.1 ENST00000431432.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008623.hg.1 ENST00000431432.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00174742.hg.1 ENST00000370503.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00174742.hg.1 NM_001009609.3 Refseq
          Affymetrix Human Clariom D Assay PSR0X00174742.hg.1 ENST00000370503.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00174742.hg.1 NM_001009609.3 Refseq
          Affymetrix Human Clariom D Assay PSR0X00174742.hg.1 TC0X00011010.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00174743.hg.1 ENST00000370503.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00174743.hg.1 NM_001009609.3 Refseq
          Affymetrix Human Clariom D Assay PSR0X00174743.hg.1 ENST00000370503.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00174743.hg.1 NM_001009609.3 Refseq
          Affymetrix Human Clariom D Assay PSR0X00174743.hg.1 TC0X00011010.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00174744.hg.1 ENST00000370503.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00174744.hg.1 NM_001009609.3 Refseq
          Affymetrix Human Clariom D Assay PSR0X00174744.hg.1 ENST00000370503.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00174744.hg.1 NM_001009609.3 Refseq
          Affymetrix Human Clariom D Assay PSR0X00174744.hg.1 TC0X00011010.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0X00011010.hg.1 ENST00000370503.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011010.hg.1 ENST00000370503.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011010.hg.1 ENST00000370503.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011010.hg.1 ENST00000370503.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011010.hg.1 NM_001009609.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00011010.hg.1 NM_001009609.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00011010.hg.1 NM_001009609.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00011010.hg.1 NM_001009609.3 Refseq
          Human Genome U133 Set 234889_PM_at - -
          Human Genome U133 Set 234368_at - -
          Human Genome U133 Set 234368_PM_at - -
          Human Genome U133 Set 234889_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A040488 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A040488 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1691953 DQ336128.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1691953 DQ336129.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1691953 NM_001009609.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1691953 BC062750.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1691953 DQ336128.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1691953 DQ336129.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1691953 NM_001009609.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1691953 BC062750.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1691953 DQ336128.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1691953 DQ336129.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1691953 NM_001009609.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1691953 BC062750.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1691953 DQ336128.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1691953 DQ336129.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1691953 NM_001009609.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1691953 BC062750.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1691953 DQ336128.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1691953 DQ336129.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1691953 NM_001009609.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1691953 BC062750.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_14551-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1691953 DQ336128.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1691953 DQ336129.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1691953 NM_001009609.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1691953 BC062750.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1691953 DQ336128.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1691953 DQ336129.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1691953 NM_001009609.2 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1691953 BC062750.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 1500349 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_14551 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1691953 DQ336128.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1691953 DQ336129.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1691953 NM_001009609.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1691953 BC062750.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1691953 DQ336128.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1691953 DQ336129.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1691953 NM_001009609.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1691953 BC062750.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1691953 DQ336128.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1691953 DQ336129.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1691953 NM_001009609.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1691953 BC062750.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1691953 DQ336128.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1691953 DQ336129.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1691953 NM_001009609.2 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1691953 BC062750.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1691953 DQ336128.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1691953 DQ336129.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1691953 NM_001009609.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1691953 BC062750.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline