| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
8071043 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8071042 |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006520.hg.1 |
ENST00000623370.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006520.hg.1 |
ENST00000623370.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000463645.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000463645.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000463645.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000463645.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000463645.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000463645.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000466372.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000466372.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000481510.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000481510.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000481510.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000623058.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000623058.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000623058.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000623058.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000623058.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000623058.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000623058.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000623058.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000623058.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000624391.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000624391.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000624391.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000624391.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000624391.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000624391.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000624391.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000624391.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
ENST00000624391.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
NM_013310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
NM_013310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
NM_013310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
NM_013310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
NM_013310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
NM_013310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009350.hg.1 |
NM_013310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006520.hg.1 |
ENST00000623370.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006520.hg.1 |
ENST00000623370.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000463645.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000463645.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000463645.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000463645.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000463645.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000463645.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000466372.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000466372.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000481510.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000481510.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000481510.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000623058.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000623058.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000623058.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000623058.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000623058.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000623058.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000623058.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000623058.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000623058.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000624391.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000624391.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000624391.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000624391.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000624391.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000624391.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000624391.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000624391.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
ENST00000624391.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
NM_013310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
NM_013310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
NM_013310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
NM_013310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
NM_013310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
NM_013310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200009350.hg.1 |
NM_013310.4 |
Refseq |