Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02761456_s1
          See other LINC01602 GE Assays ›
          Gene Symbol:
          LINC01602
          Gene Name:
          long intergenic non-protein coding RNA 1602
          Gene Aliases:
          T1560
          Chromosome Location:
          Chr.8: 57978358 - 57984126 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02761456_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC01602
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_130934.1 3 1419 146
          GenBank mRNA BC040611.2 - 3 1343 146

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800007729.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427503 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06143910 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427495 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427487 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06143967 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427530 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427525 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427527 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427541 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06143952 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427498 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427526 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427484 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427517 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06143932 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06143919 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06143920 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427522 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06143923 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427540 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427528 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427533 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427486 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427500 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06143926 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427532 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427488 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427494 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427545 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06143915 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427513 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427539 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427546 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427505 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427518 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427485 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427509 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427516 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427538 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427515 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427537 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427490 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06143913 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427534 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427523 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06143945 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427531 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427483 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06143970 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06143925 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427510 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427514 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427482 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427543 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26427548 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3397579 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 26818 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3397579 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 19647 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 71820 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P16498028 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18316667 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P32271985 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8146531 - -
          Affymetrix Human Gene 1.0 ST Array 8146528 - -
          Affymetrix Human Gene 1.0 ST Array 8146530 - -
          Affymetrix Human Gene 1.0 ST Array 8146532 - -
          Affymetrix Human Gene 1.0 ST Array 8146529 - -
          Affymetrix Human Gene 1.1 ST Array 8146527 - -
          Affymetrix Human Transcriptome Array 2.0 TC0800007729.hg.1 ENST00000522992.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007729.hg.1 ENST00000522992.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007729.hg.1 ENST00000522992.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007729.hg.1 NR_130934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007729.hg.1 NR_130934.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007729.hg.1 NR_130934.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800153171.hg.1 ENST00000522992.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800153171.hg.1 NR_130934.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800153171.hg.1 ENST00000522992.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800153171.hg.1 NR_130934.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800153171.hg.1 TC0800007729.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800153170.hg.1 ENST00000522992.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800153170.hg.1 NR_130934.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800153170.hg.1 ENST00000522992.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800153170.hg.1 NR_130934.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800153170.hg.1 TC0800007729.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0800007729.hg.1 ENST00000522992.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007729.hg.1 ENST00000522992.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007729.hg.1 ENST00000522992.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007729.hg.1 NR_130934.1 Refseq
          Affymetrix Human Clariom D Assay TC0800007729.hg.1 NR_130934.1 Refseq
          Affymetrix Human Clariom D Assay TC0800007729.hg.1 NR_130934.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800153169.hg.1 ENST00000522992.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800153169.hg.1 NR_130934.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800153169.hg.1 ENST00000522992.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800153169.hg.1 NR_130934.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800153169.hg.1 TC0800007729.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline