| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
NM_033061_at |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8019570 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8019699 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8015218 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8007112 |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
216810_PM_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11742373_s_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11742373_PM_s_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006056.hg.1 |
ENST00000391417.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006056.hg.1 |
NM_033061.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006056.hg.1 |
TC17000502.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006055.hg.1 |
ENST00000377734.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006055.hg.1 |
ENST00000391417.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006055.hg.1 |
NM_033061.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006055.hg.1 |
TC17000502.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012251.hg.1 |
ENST00000391417.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012251.hg.1 |
ENST00000391417.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012251.hg.1 |
ENST00000391417.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012251.hg.1 |
ENST00000391417.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012251.hg.1 |
ENST00000391417.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012251.hg.1 |
ENST00000621138.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012251.hg.1 |
ENST00000621138.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012251.hg.1 |
NM_033061.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012251.hg.1 |
NM_033061.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012251.hg.1 |
NM_033061.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012251.hg.1 |
NM_033061.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
ENST00000318329.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
ENST00000318329.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
ENST00000318329.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012252.hg.1 |
ENST00000391415.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012252.hg.1 |
ENST00000391415.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012252.hg.1 |
ENST00000391415.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012252.hg.1 |
ENST00000391415.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012252.hg.1 |
ENST00000391415.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012252.hg.1 |
ENST00000391415.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012252.hg.1 |
ENST00000391415.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012252.hg.1 |
ENST00000617453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012252.hg.1 |
ENST00000617453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012252.hg.1 |
ENST00000617453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012252.hg.1 |
NM_001146041.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012252.hg.1 |
NM_001146041.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012252.hg.1 |
NM_001146041.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012252.hg.1 |
NM_001146041.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012252.hg.1 |
NM_001146041.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012252.hg.1 |
NM_001146041.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006057.hg.1 |
ENST00000377734.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006057.hg.1 |
ENST00000391417.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006057.hg.1 |
NM_033061.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006057.hg.1 |
TC17000502.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006059.hg.1 |
ENST00000391417.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006059.hg.1 |
NM_033061.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006059.hg.1 |
TC17000502.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1700202321.hg.1 |
ENST00000391417.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700202321.hg.1 |
ENST00000621138.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700202321.hg.1 |
NM_033061.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700202321.hg.1 |
ENST00000391417.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700202321.hg.1 |
ENST00000621138.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700202321.hg.1 |
NM_033061.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700202321.hg.1 |
TC1700012251.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700202323.hg.1 |
ENST00000391417.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700202323.hg.1 |
ENST00000621138.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700202323.hg.1 |
NM_033061.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700202323.hg.1 |
ENST00000391417.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700202323.hg.1 |
ENST00000621138.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700202323.hg.1 |
NM_033061.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700202323.hg.1 |
TC1700012251.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700202324.hg.1 |
ENST00000391417.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700202324.hg.1 |
NM_033061.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700202324.hg.1 |
ENST00000391417.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700202324.hg.1 |
NM_033061.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700202324.hg.1 |
TC1700012251.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
ENST00000318329.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
ENST00000318329.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
ENST00000318329.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012251.hg.1 |
ENST00000391417.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012251.hg.1 |
ENST00000391417.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012251.hg.1 |
ENST00000391417.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012251.hg.1 |
ENST00000391417.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012251.hg.1 |
ENST00000391417.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012251.hg.1 |
ENST00000621138.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012251.hg.1 |
ENST00000621138.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012251.hg.1 |
NM_033061.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012251.hg.1 |
NM_033061.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012251.hg.1 |
NM_033061.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012251.hg.1 |
NM_033061.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700202322.hg.1 |
ENST00000391417.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700202322.hg.1 |
NM_033061.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700202322.hg.1 |
ENST00000391417.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700202322.hg.1 |
NM_033061.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700202322.hg.1 |
TC1700012251.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1700012252.hg.1 |
ENST00000391415.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012252.hg.1 |
ENST00000391415.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012252.hg.1 |
ENST00000391415.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012252.hg.1 |
ENST00000391415.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012252.hg.1 |
ENST00000391415.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012252.hg.1 |
ENST00000391415.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012252.hg.1 |
ENST00000391415.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012252.hg.1 |
ENST00000617453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012252.hg.1 |
ENST00000617453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012252.hg.1 |
ENST00000617453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012252.hg.1 |
NM_001146041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012252.hg.1 |
NM_001146041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012252.hg.1 |
NM_001146041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012252.hg.1 |
NM_001146041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012252.hg.1 |
NM_001146041.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012252.hg.1 |
NM_001146041.1 |
Refseq |
| Human Genome U133 Plus 2.0 Array |
216810_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
216810_PM_at |
- |
- |