| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000371419.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000371419.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000371419.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000371419.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000371419.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000371419.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000371419.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000371419.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000371419.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000441080.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000441080.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000441080.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000441080.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000441080.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000441080.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000474326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000474326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000487129.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000487129.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000487129.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000487129.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000487129.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000487129.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000487129.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000487129.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000493356.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000493356.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000493356.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000493356.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000493356.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000493356.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
ENST00000493356.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
NM_002623.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
NM_002623.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
NM_002623.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
NM_002623.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
NM_002623.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
NM_002623.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
NM_002623.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007792.hg.1 |
NM_002623.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008243.hg.1 |
ENST00000582071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000371419.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000371419.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000371419.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000371419.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000371419.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000371419.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000371419.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000371419.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000371419.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000441080.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000441080.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000441080.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000441080.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000441080.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000441080.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000474326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000474326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000487129.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000487129.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000487129.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000487129.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000487129.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000487129.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000487129.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000487129.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000493356.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000493356.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000493356.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000493356.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000493356.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000493356.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
ENST00000493356.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
NM_002623.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
NM_002623.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
NM_002623.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
NM_002623.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
NM_002623.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
NM_002623.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
NM_002623.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007792.hg.1 |
NM_002623.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008243.hg.1 |
ENST00000582071.1 |
Ensembl |