Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03003820_m1
          See other ZC3H18-AS1 GE Assays ›
          Gene Symbol:
          ZC3H18-AS1
          Gene Name:
          ZC3H18 antisense RNA 1 (head to head)
          Gene Aliases:
          C16orf85, LINC00859
          Chromosome Location:
          Chr.16: 88553981 - 88567106 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03003820_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ZC3H18-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK127438.1 - 2 - 3 2826 95

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178561 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178611 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30723739 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30723738 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178545 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178566 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178560 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178585 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178575 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178618 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178576 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30723767 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178556 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178573 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16927693 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178612 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178626 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178625 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178627 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178553 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178557 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30723812 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178582 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178621 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178596 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16927705 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30723827 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30723820 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178599 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30723763 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178546 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178564 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178583 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178606 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178571 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178567 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16927694 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178558 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30723817 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178579 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178589 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30723821 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178622 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178616 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30723731 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178603 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178588 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30723780 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178623 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178581 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178597 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30723816 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30723781 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178577 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178559 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178610 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16927691 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178551 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178594 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178590 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178570 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178598 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30723819 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178592 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178568 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178565 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30723807 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178554 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178607 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178593 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178574 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178605 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178547 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178604 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178572 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10178544 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3398111 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0128 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0127 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0126 - -
          Agilent Human Genome CGH Microarray 44B A_14_P117952 - -
          Agilent Human Genome CGH Microarray 44A A_14_P117952 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 46982 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P03197494 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P117952 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10178623 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10178616 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10178614 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10178626 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10178619 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10178621 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10178628 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10178620 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P10178626 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16927709 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P10178628 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P10178627 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16927708 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P10178625 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16927710 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16927700 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03197508 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03197494 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P117952 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 169213 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 99666 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 97891 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 59876 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P33995046 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P117952 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03197496 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03197501 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03197508 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20556654 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03197494 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P33995076 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03197506 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001001682_at - -
          Affymetrix Human Gene 1.0 ST Array 8003320 - -
          Affymetrix Human Gene 1.0 ST Array 8003321 - -
          Affymetrix Human Gene 1.0 ST Array 8003319 - -
          Affymetrix Human Gene 1.1 ST Array 8003318 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1806657 AK127438.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1806657 AK127438.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1806657 AK127438.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1806657 AK127438.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1806657 AK127438.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_29616-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1806657 AK127438.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1806657 AK127438.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip ILMN_29616 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_29616 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1806657 AK127438.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1806657 AK127438.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1806657 AK127438.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1806657 AK127438.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1806657 AK127438.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0