| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
8104140 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8104139 |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
226558_PM_at |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
244631_PM_at |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
242577_PM_at |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
241624_PM_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11746708_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11746708_PM_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007536.hg.1 |
ENST00000624968.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TCUn_GL000218v100006433.hg.1 |
ENST00000612565.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TCUn_GL000218v100006433.hg.1 |
NR_027420.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TCUn_GL000218v100006433.hg.1 |
NR_027420.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TCUn_GL000218v100006433.hg.1 |
NR_027420.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000400783.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000400783.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000400783.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000400783.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000568768.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000568768.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000568768.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000568768.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000568768.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000568768.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000568768.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000568768.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000568768.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000568768.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000568768.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000582835.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000582835.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000582835.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000582835.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000582835.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000582835.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000582835.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000582835.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000582835.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000582835.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000582835.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000582835.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
ENST00000582835.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
NR_037597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
NR_037597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
NR_037597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
NR_037597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
NR_037597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
NR_037597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
NR_037597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
NR_037597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
NR_037597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
NR_037597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
NR_037597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
NR_037597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013456.hg.1 |
NR_037597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008445.hg.1 |
ENST00000618617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSRUn_GL000218v100144048.hg.1 |
TCUn_GL000218v100006433.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSRUn_GL000218v100144048.hg.1 |
ENST00000612565.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSRUn_GL000218v100144048.hg.1 |
NR_027420.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSRUn_GL000218v100144048.hg.1 |
ENST00000612565.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSRUn_GL000218v100144048.hg.1 |
NR_027420.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSRUn_GL000218v100144047.hg.1 |
TCUn_GL000218v100006433.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSRUn_GL000218v100144047.hg.1 |
NR_027420.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSRUn_GL000218v100144047.hg.1 |
NR_027420.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSRUn_GL000218v100144049.hg.1 |
TCUn_GL000218v100006433.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSRUn_GL000218v100144049.hg.1 |
NR_027420.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSRUn_GL000218v100144049.hg.1 |
NR_027420.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000400783.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000400783.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000400783.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000400783.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000568768.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000568768.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000568768.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000568768.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000568768.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000568768.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000568768.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000568768.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000568768.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000568768.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000568768.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000582835.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000582835.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000582835.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000582835.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000582835.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000582835.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000582835.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000582835.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000582835.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000582835.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000582835.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000582835.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
ENST00000582835.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
NR_037597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
NR_037597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
NR_037597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
NR_037597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
NR_037597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
NR_037597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
NR_037597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
NR_037597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
NR_037597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
NR_037597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
NR_037597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
NR_037597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013456.hg.1 |
NR_037597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TCUn_GL000218v100006433.hg.1 |
ENST00000612565.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TCUn_GL000218v100006433.hg.1 |
NR_027420.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TCUn_GL000218v100006433.hg.1 |
NR_027420.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TCUn_GL000218v100006433.hg.1 |
NR_027420.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008445.hg.1 |
ENST00000618617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007536.hg.1 |
ENST00000624968.1 |
Ensembl |
| Human Genome U133 Plus 2.0 Array |
244631_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
244631_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
241624_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
241624_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
241624_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
241624_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
244631_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
241624_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
244631_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
241624_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
241624_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
244631_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
241624_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
244631_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
241624_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
244631_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
244631_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
244631_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
241624_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
241624_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
241624_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
244631_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
244631_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
241624_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
244631_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
244631_at3 |
- |
- |
| Human Genome U95 Set |
73422_PM_at |
- |
- |
| Human Genome U95 Set |
73422_at |
- |
- |
| Human Genome U95 Set |
85497_at |
- |
- |
| Human Genome U95 Set |
85497_PM_at |
- |
- |
| Human Genome U133 Set |
241624_at |
- |
- |
| Human Genome U133 Set |
244631_PM_at |
- |
- |
| Human Genome U133 Set |
241625_at |
- |
- |
| Human Genome U133 Set |
241624_PM_at |
- |
- |
| Human Genome U133 Set |
241625_PM_at |
- |
- |
| Human Genome U133 Set |
244631_at |
- |
- |