Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03026365_s1
          See other DEFB115 GE Assays ›
          Gene Symbol:
          DEFB115
          Gene Name:
          defensin beta 115
          Gene Aliases:
          DEFB-15
          Chromosome Location:
          Chr.20: 31257664 - 31259632 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03026365_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DEFB115
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037730.1 NP_001032819.1 2 196 147
          GenBank mRNA DQ012019.1 - 2 196 147

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000007064.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000181.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Inventoried Amplicon spans exons and probe does not span exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DEFB115

          Panther Classification:

          Molecular Function -

          antimicrobial response protein defense/immunity protein

          Gene Ontology Categories:

          Function(s)

          defense response to bacterium
          innate immune response

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31776798 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31776788 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31776742 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31776748 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11129682 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31776790 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31776772 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31776796 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11129687 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11129691 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11129715 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11129685 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31776787 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31776773 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17186286 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31776752 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31776780 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31776769 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31776767 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31776792 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11129680 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31776750 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11129702 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11129714 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31776779 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11129686 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11129694 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11129710 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31776765 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11129689 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31776764 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31776771 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31776760 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11129678 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11129709 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11129690 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31776751 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11129697 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11129708 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31776768 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31776793 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11129673 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11129711 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11129681 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31776741 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11129730 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31776774 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11129684 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31776786 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 132690 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 277973 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 79158 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3328352 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1243 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1244 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 10743 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P21110147 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P13774517 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3328352 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3328352 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11129694 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31776760 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11129674 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11129691 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31776748 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31776768 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11129681 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11129684 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31776742 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31776767 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11129673 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31776764 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11129682 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11129680 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11129697 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11129686 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11129687 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31776750 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31776751 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31776745 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31776769 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11129702 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11129697 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11129684 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11129686 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31776742 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31776751 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11129681 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11129691 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11129702 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11129673 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11129694 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11129687 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31776748 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31776768 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31776760 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11129680 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31776764 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31776767 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11129674 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31776769 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31776745 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31776750 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11129682 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P21110147 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P21110147 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 89472 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 74360 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P13777531 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P21110147 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11129693 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11129679 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11129708 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11129670 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11129684 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11129711 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11129688 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11129677 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11129700 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11129714 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11129697 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11129674 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11129716 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11129722 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11129705 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11129703 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11129672 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11129681 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11129730 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11129727 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17186288 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11129691 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11129695 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11129698 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11129718 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11129713 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P21110147 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001037730_at - -
          Affymetrix Human Gene 1.0 ST Array 8061506 - -
          Affymetrix Human Gene 1.0 ST Array 8061505 - -
          Affymetrix Human Gene 1.1 ST Array 8061504 - -
          Affymetrix Human Genome U219 Array 11738973_PM_at - -
          Affymetrix Human Genome U219 Array 11738973_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR20002235.hg.1 ENST00000400552.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20002235.hg.1 NM_001037730.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20002235.hg.1 TC20000181.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC2000007064.hg.1 ENST00000400552.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007064.hg.1 ENST00000400552.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007064.hg.1 NM_001037730.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007064.hg.1 NM_001037730.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20002234.hg.1 ENST00000400552.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20002234.hg.1 NM_001037730.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20002234.hg.1 TC20000181.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC2000007064.hg.1 ENST00000400552.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000007064.hg.1 ENST00000400552.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000007064.hg.1 NM_001037730.1 Refseq
          Affymetrix Human Clariom D Assay TC2000007064.hg.1 NM_001037730.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000148203.hg.1 ENST00000400552.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148203.hg.1 NM_001037730.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000148203.hg.1 TC2000007064.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000148203.hg.1 ENST00000400552.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148203.hg.1 NM_001037730.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000148204.hg.1 ENST00000400552.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148204.hg.1 NM_001037730.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000148204.hg.1 TC2000007064.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000148204.hg.1 ENST00000400552.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148204.hg.1 NM_001037730.1 Refseq
          Human Genome U133 Set 234451_PM_at - -
          Human Genome U133 Set 234451_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A018173 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A018173 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3248525 NM_001037730.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3248525 DQ012019.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3248525 NM_001037730.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3248525 DQ012019.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3248525 NM_001037730.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3248525 DQ012019.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3248525 NM_001037730.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3248525 DQ012019.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline