Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03026654_u1
          See other LOC650293 GE Assays ›
          Gene Symbol:
          LOC650293
          Gene Name:
          seven transmembrane helix receptor
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 8949751 - 8950401 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03026654_u1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC650293
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001040071.1 NP_001035160.1 1 574 134

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0400006773.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC04000093.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23070816 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 800787 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 74586 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3405535 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P352358 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 44395 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 20950 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 51500 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P352358 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3405535 - -
          SurePrint G3 Human GE Microarray G4851A A_32_P352358 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3405535 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P352358 - -
          Agilent Human 1B Microarray G4111A A_32_P427731 - -
          Agilent Human 1B Microarray G4111A A_32_P456046 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02919516 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15543956 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7960899 - -
          Affymetrix Human Gene 1.0 ST Array 8149149 - -
          Affymetrix Human Gene 1.0 ST Array 8144497 - -
          Affymetrix Human Gene 1.0 ST Array 7960897 - -
          Affymetrix Human Gene 1.0 ST Array 8149209 - -
          Affymetrix Human Gene 1.0 ST Array 8144423 - -
          Affymetrix Human Gene 1.0 ST Array 7949897 - -
          Affymetrix Human Genome U219 Array 11715160_PM_x_at - -
          Affymetrix Human Genome U219 Array 11715160_x_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR04001965.hg.1 TC04000093.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04001965.hg.1 NM_001040071.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 ENST00000309950.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 ENST00000309950.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 ENST00000309950.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 ENST00000309950.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 ENST00000309950.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 ENST00000309950.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 ENST00000309950.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 ENST00000309950.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 ENST00000409042.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 ENST00000409042.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 ENST00000409042.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 ENST00000409042.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 ENST00000409042.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 ENST00000409042.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 ENST00000409042.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 ENST00000411729.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 ENST00000411729.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 ENST00000621326.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 ENST00000621326.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 ENST00000621326.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 ENST00000621326.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 ENST00000621326.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 ENST00000621326.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 ENST00000621326.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 ENST00000621326.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 ENST00000621326.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 NM_001017920.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 NM_001017920.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 NM_001017920.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 NM_001017920.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 NM_001017920.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 NM_001017920.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 NM_001017920.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016553.hg.1 NM_001017920.2 Refseq
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 ENST00000309950.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 ENST00000309950.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 ENST00000309950.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 ENST00000309950.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 ENST00000309950.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 ENST00000309950.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 ENST00000309950.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 ENST00000309950.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 ENST00000409042.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 ENST00000409042.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 ENST00000409042.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 ENST00000409042.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 ENST00000409042.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 ENST00000409042.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 ENST00000409042.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 ENST00000411729.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 ENST00000411729.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 ENST00000621326.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 ENST00000621326.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 ENST00000621326.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 ENST00000621326.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 ENST00000621326.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 ENST00000621326.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 ENST00000621326.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 ENST00000621326.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 ENST00000621326.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 NM_001017920.2 Refseq
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 NM_001017920.2 Refseq
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 NM_001017920.2 Refseq
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 NM_001017920.2 Refseq
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 NM_001017920.2 Refseq
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 NM_001017920.2 Refseq
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 NM_001017920.2 Refseq
          Affymetrix Human Clariom D Assay TC0200016553.hg.1 NM_001017920.2 Refseq
          Affymetrix Human Clariom D Assay PSR0400147007.hg.1 ENST00000508957.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400147007.hg.1 NM_001040071.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400147007.hg.1 ENST00000508957.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400147007.hg.1 NM_001040071.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400147007.hg.1 TC0400006773.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 216476_PM_at - -
          Human Genome U133 Plus 2.0 Array 215770_PM_at - -
          Human Genome U133 Plus 2.0 Array 215770_at - -
          Human Genome U133 Plus 2.0 Array 216476_at - -
          Human Genome U133A 2.0 Array 215770_PM_at - -
          Human Genome U133A 2.0 Array 215770_at - -
          Human Genome U133A 2.0 Array 216476_PM_at - -
          Human Genome U133A 2.0 Array 216476_at - -
          Human Genome U133 Set 215770_PM_at - -
          Human Genome U133 Set 216476_PM_at - -
          Human Genome U133 Set 215770_at - -
          Human Genome U133 Set 216476_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A025257 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A025257 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1685813 - -
          HumanHT-12 v4 Expression BeadChip ILMN_2079225 NM_001040071.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2079225 AW009208.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1685813 - -
          HumanHT-12 V4.0 expression beadchip ILMN_2079225 NM_001040071.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2079225 AW009208.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2079225 NM_001040071.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2079225 AW009208.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2079225 NM_001040071.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2079225 AW009208.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1685813 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2079225 NM_001040071.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2079225 AW009208.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1685813 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2079225 NM_001040071.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2079225 AW009208.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2079225 NM_001040071.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2079225 AW009208.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1685813 - -
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2079225 NM_001040071.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2079225 AW009208.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0