Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03037724_m1
          See other LRRC14B GE Assays ›
          Gene Symbol:
          LRRC14B
          Gene Name:
          leucine rich repeat containing 14B
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 191511 - 195353 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03037724_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LRRC14B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001080478.1 NP_001073947.1 1 - 2 926 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500006437.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC05000003.hg.1

          Set Membership:

          Amplicon length between 71 and 85 Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03708960 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03708955 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03708966 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03708954 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03708962 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03708985 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03708956 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15651825 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03708987 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15651797 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03708968 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23872331 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03708949 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03708973 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03708958 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03708971 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03708980 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23872358 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23872373 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23872334 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03708950 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23872378 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03708959 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23872369 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23872343 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23872315 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03708978 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03708953 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03708981 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03708969 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03708974 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03708982 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23872307 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23872368 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23872351 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23872335 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 37829 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 420545 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 241663 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 263090 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3253538 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1450 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1451 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 61669 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P01163023 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P15059100 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P01163023 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3253538 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3253538 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03708956 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03708953 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23872331 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03708955 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23872307 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03708958 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03708949 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03708948 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03708962 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03708960 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23872334 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03708950 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23872335 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03708958 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03708950 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03708948 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03708956 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23872307 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23872334 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03708953 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23872331 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03708955 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23872335 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03708949 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03708962 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03708960 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 86966 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 73577 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01163029 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01163023 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708976 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708981 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708971 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708992 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708964 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708948 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15651826 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15651790 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15651792 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15651825 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708974 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708956 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15651804 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708960 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708952 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708990 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708977 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708988 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708973 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15651819 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708982 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708968 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708955 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708969 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708963 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708966 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708985 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708993 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708980 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708978 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708950 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708958 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708987 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708994 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03708991 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P01163029 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001080478_at - -
          Affymetrix Human Gene 1.0 ST Array 8104165 - -
          Affymetrix Human Gene 1.0 ST Array 8104164 - -
          Affymetrix Human Gene 1.1 ST Array 8104163 - -
          Affymetrix Human Genome U219 Array 11741740_at - -
          Affymetrix Human Genome U219 Array 11741740_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR05000031.hg.1 TC05000003.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05000031.hg.1 ENST00000328278.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05000031.hg.1 NM_001080478.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05000030.hg.1 TC05000003.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05000030.hg.1 ENST00000328278.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05000030.hg.1 NM_001080478.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05000032.hg.1 TC05000003.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05000032.hg.1 ENST00000328278.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05000032.hg.1 NM_001080478.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500006437.hg.1 ENST00000328278.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500006437.hg.1 ENST00000328278.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500006437.hg.1 ENST00000328278.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500006437.hg.1 NM_001080478.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500006437.hg.1 NM_001080478.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500006437.hg.1 NM_001080478.2 Refseq
          Affymetrix Human Clariom D Assay TC0500006437.hg.1 ENST00000328278.3 Ensembl
          Affymetrix Human Clariom D Assay TC0500006437.hg.1 ENST00000328278.3 Ensembl
          Affymetrix Human Clariom D Assay TC0500006437.hg.1 ENST00000328278.3 Ensembl
          Affymetrix Human Clariom D Assay TC0500006437.hg.1 NM_001080478.2 Refseq
          Affymetrix Human Clariom D Assay TC0500006437.hg.1 NM_001080478.2 Refseq
          Affymetrix Human Clariom D Assay TC0500006437.hg.1 NM_001080478.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500144091.hg.1 ENST00000328278.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0500144091.hg.1 NM_001080478.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500144091.hg.1 ENST00000328278.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0500144091.hg.1 NM_001080478.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500144091.hg.1 TC0500006437.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500144093.hg.1 ENST00000328278.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0500144093.hg.1 NM_001080478.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500144093.hg.1 ENST00000328278.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0500144093.hg.1 NM_001080478.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500144093.hg.1 TC0500006437.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500144094.hg.1 ENST00000328278.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0500144094.hg.1 NM_001080478.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500144094.hg.1 ENST00000328278.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0500144094.hg.1 NM_001080478.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500144094.hg.1 TC0500006437.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A027834 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A027834 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 204449 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_2241252 NM_001080478.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1796264 - -
          HumanHT-12 V4.0 expression beadchip ILMN_2241252 NM_001080478.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1814647 NM_001080478.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2241252 NM_001080478.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2241252 NM_001080478.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1796264 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2241252 NM_001080478.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1814647 NM_001080478.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1814647 NM_001080478.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2241252 NM_001080478.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1814647 NM_001080478.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2241252 NM_001080478.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1796264 - -
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1814647 NM_001080478.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2241252 NM_001080478.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline