Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03044337_g1
          See other C7ORF34 GE Assays ›
          Gene Symbol:
          C7orf34
          Gene Name:
          chromosome 7 open reading frame 34
          Gene Aliases:
          ctm-1
          Chromosome Location:
          Chr.7: 142939506 - 142940870 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03044337_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C7orf34
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178829.4 NP_849151.2 1 - 2 251 69
          GenBank mRNA AB083484.1 - 1 - 2 199 69
          BC014596.1 - 1 - 2 256 4825428 69
          KU178863.1 - 1 - 2 135 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700009478.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07000925.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene C7orf34

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR07014308.hg.1 ENST00000409607.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07014308.hg.1 NM_178829.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07014308.hg.1 TC07000925.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0700009478.hg.1 ENST00000409607.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009478.hg.1 ENST00000409607.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009478.hg.1 ENST00000409607.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009478.hg.1 ENST00000409607.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009478.hg.1 ENST00000409607.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009478.hg.1 ENST00000458732.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009478.hg.1 ENST00000458732.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009478.hg.1 ENST00000458732.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009478.hg.1 ENST00000458732.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009478.hg.1 NM_178829.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700009478.hg.1 NM_178829.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700009478.hg.1 NM_178829.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700009478.hg.1 NM_178829.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700009478.hg.1 NM_178829.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07014309.hg.1 ENST00000409607.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07014309.hg.1 NM_178829.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07014309.hg.1 TC07000925.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07014310.hg.1 ENST00000409607.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07014310.hg.1 NM_178829.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07014310.hg.1 TC07000925.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0700167308.hg.1 ENST00000409607.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0700167308.hg.1 ENST00000458732.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700167308.hg.1 NM_178829.4 Refseq
          Affymetrix Human Clariom D Assay PSR0700167308.hg.1 ENST00000409607.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0700167308.hg.1 ENST00000458732.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700167308.hg.1 NM_178829.4 Refseq
          Affymetrix Human Clariom D Assay PSR0700167308.hg.1 TC0700009478.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700167309.hg.1 ENST00000409607.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0700167309.hg.1 ENST00000458732.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700167309.hg.1 NM_178829.4 Refseq
          Affymetrix Human Clariom D Assay PSR0700167309.hg.1 ENST00000409607.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0700167309.hg.1 ENST00000458732.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700167309.hg.1 NM_178829.4 Refseq
          Affymetrix Human Clariom D Assay PSR0700167309.hg.1 TC0700009478.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700167306.hg.1 ENST00000409607.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0700167306.hg.1 NM_178829.4 Refseq
          Affymetrix Human Clariom D Assay PSR0700167306.hg.1 ENST00000409607.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0700167306.hg.1 NM_178829.4 Refseq
          Affymetrix Human Clariom D Assay PSR0700167306.hg.1 TC0700009478.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700167307.hg.1 ENST00000409607.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0700167307.hg.1 ENST00000458732.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700167307.hg.1 NM_178829.4 Refseq
          Affymetrix Human Clariom D Assay PSR0700167307.hg.1 ENST00000409607.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0700167307.hg.1 ENST00000458732.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700167307.hg.1 NM_178829.4 Refseq
          Affymetrix Human Clariom D Assay PSR0700167307.hg.1 TC0700009478.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700167305.hg.1 ENST00000409607.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0700167305.hg.1 NM_178829.4 Refseq
          Affymetrix Human Clariom D Assay PSR0700167305.hg.1 ENST00000409607.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0700167305.hg.1 NM_178829.4 Refseq
          Affymetrix Human Clariom D Assay PSR0700167305.hg.1 TC0700009478.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0700009478.hg.1 ENST00000409607.3 Ensembl
          Affymetrix Human Clariom D Assay TC0700009478.hg.1 ENST00000409607.3 Ensembl
          Affymetrix Human Clariom D Assay TC0700009478.hg.1 ENST00000409607.3 Ensembl
          Affymetrix Human Clariom D Assay TC0700009478.hg.1 ENST00000409607.3 Ensembl
          Affymetrix Human Clariom D Assay TC0700009478.hg.1 ENST00000409607.3 Ensembl
          Affymetrix Human Clariom D Assay TC0700009478.hg.1 ENST00000458732.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009478.hg.1 ENST00000458732.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009478.hg.1 ENST00000458732.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009478.hg.1 ENST00000458732.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009478.hg.1 NM_178829.4 Refseq
          Affymetrix Human Clariom D Assay TC0700009478.hg.1 NM_178829.4 Refseq
          Affymetrix Human Clariom D Assay TC0700009478.hg.1 NM_178829.4 Refseq
          Affymetrix Human Clariom D Assay TC0700009478.hg.1 NM_178829.4 Refseq
          Affymetrix Human Clariom D Assay TC0700009478.hg.1 NM_178829.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 145879 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline