| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20571850 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15129621 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20571843 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00514800 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00514696 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20572046 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20572067 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00514685 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20571860 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20571864 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20572047 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15129617 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00514799 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20571865 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20571851 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20572057 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20571867 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00514694 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20572052 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20571831 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20571855 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20572048 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20571876 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00514820 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20571866 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00514812 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20571848 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20571842 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15129734 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20572064 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20572070 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20571874 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00514690 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20571880 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20572054 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00514796 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15129728 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20571844 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20571840 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15129620 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00514703 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00514686 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20571854 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00514688 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00514701 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20571875 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20572051 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20571879 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00514693 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00514811 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00514802 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00514809 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20571858 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20571870 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20572061 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00514803 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20571873 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20572044 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20572063 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20571853 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15129604 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
175892 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
12643 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
934662 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
772269 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
552069 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
40646 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
231847 |
AY177732.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
231847 |
BC057760.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
231847 |
CR610648.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
231847 |
NM_173855.3 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
576897 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
731754 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
690017 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3260426 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3260430 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
35149 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
49333 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
32569 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
35575 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_18_P10325214 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3260430 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3260426 |
- |
- |
| SurePrint G3 Human GE Microarray G4851A |
A_33_P3260426 |
- |
- |
| SurePrint G3 Human GE Microarray G4851A |
A_19_P00315999 |
- |
- |
| SurePrint G3 Human GE Microarray G4851A |
A_33_P3260430 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571879 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571866 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15129591 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571855 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571854 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571843 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571831 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571864 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571851 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571880 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15129604 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00514690 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571840 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00514694 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15129617 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571870 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571873 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571874 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571842 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571875 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15129620 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00514693 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15129621 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00514688 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571867 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571865 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571876 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571860 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15129600 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00514701 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571858 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571848 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571853 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00514686 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00514688 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571864 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15129591 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15129617 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00514690 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15129620 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571865 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00514701 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571851 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571867 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571843 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571842 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571880 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15129604 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571853 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571855 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571876 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571879 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571840 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00514693 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00514694 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571873 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571860 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571854 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571875 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571866 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571848 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571874 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15129600 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15129621 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00514686 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571870 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571831 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20571858 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00514682 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00514689 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00514704 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00514706 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00514681 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00514684 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00514680 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00514694 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00514691 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00514687 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00514700 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00514686 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00514693 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00514696 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00514702 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00514690 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00514697 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00514707 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00514701 |
- |
- |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
45025 |
- |
- |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
27653 |
- |
- |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
76303 |
- |
- |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
179309 |
- |
- |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
101698 |
- |
- |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
16493 |
- |
- |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
85027 |
- |
- |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
79913 |
- |
- |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
64148 |
- |
- |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
19022 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P15297835 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P00165914 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P17843790 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P00165957 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P10328932 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P00165940 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P00165904 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P35310670 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P15297917 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P00165925 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P15297818 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P15297889 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P35310630 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P15297920 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P15297902 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P15297903 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P15297906 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P00165921 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P00165954 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P35310653 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P10327392 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P17843074 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P106802 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P35310600 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P00165959 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P10328294 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15129600 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00514681 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00514696 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00514689 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00514692 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00514702 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00514690 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00514704 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00514686 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15129604 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00514682 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00514706 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00514700 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00514701 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00514707 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00514697 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15129618 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P00165957 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P15297903 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P15297917 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P35310670 |
- |
- |