Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03054740_sH
          See other TAS2R30 GE Assays ›
          Gene Symbol:
          TAS2R30
          Gene Name:
          taste 2 receptor member 30
          Gene Aliases:
          T2R30, T2R47, TAS2R47
          Chromosome Location:
          Chr.12: 11133285 - 11134244 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03054740_sH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TAS2R30
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001097643.1 NP_001091112.1 1 774 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200012763.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12001239.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of chemical stimulus involved in sensory perception of bitter taste
          G-protein coupled receptor signaling pathway
          G-protein coupled receptor activity
          bitter taste receptor activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28623220 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16533658 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28623196 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28623237 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28623208 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28623239 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28623228 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28623244 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16533608 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28623219 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28623210 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28623218 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28623186 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28623216 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28623189 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16533638 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28623223 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16533671 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28623214 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28623222 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28623192 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28623238 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28623231 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3294951 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 38839 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3294951 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3294951 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16533608 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623240 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623210 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623237 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623224 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623205 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623234 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623229 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623203 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623231 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623225 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16533638 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623189 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16533658 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623228 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623216 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623244 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623232 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623192 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623207 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623213 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623243 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623239 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623186 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623233 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623222 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623201 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623196 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623219 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16533671 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623198 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623218 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623220 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623222 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623228 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623189 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623224 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623210 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623218 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623243 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623207 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623220 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623244 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623186 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623192 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623213 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623239 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623198 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16533638 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623225 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623231 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16533608 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623196 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623237 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623216 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623234 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623201 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623233 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623240 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16533658 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623229 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623219 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623232 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623205 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16533671 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28623203 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Genome U219 Array 11715255_PM_s_at - -
          Affymetrix Human Genome U219 Array 11715255_s_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR12016279.hg.1 NM_001097643.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12016279.hg.1 TC12001239.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 ENST00000531678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 ENST00000531678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 ENST00000531678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 ENST00000539585.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 ENST00000539585.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 ENST00000539585.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 ENST00000539585.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 ENST00000539585.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 ENST00000541175.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 ENST00000541175.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 ENST00000541175.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 ENST00000610639.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 ENST00000610639.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 NM_001097643.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 NM_001097643.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 NM_001097643.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 NM_176884.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 NM_176884.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 NM_176884.2 Refseq
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 ENST00000531678.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 ENST00000531678.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 ENST00000531678.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 ENST00000539585.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 ENST00000539585.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 ENST00000539585.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 ENST00000539585.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 ENST00000539585.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 ENST00000541175.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 ENST00000541175.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 ENST00000541175.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 ENST00000610639.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 ENST00000610639.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 NM_001097643.1 Refseq
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 NM_001097643.1 Refseq
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 NM_001097643.1 Refseq
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 NM_176884.2 Refseq
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 NM_176884.2 Refseq
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 NM_176884.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200202700.hg.1 ENST00000539585.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200202700.hg.1 ENST00000610639.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200202700.hg.1 NM_001097643.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200202700.hg.1 ENST00000539585.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200202700.hg.1 ENST00000610639.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200202700.hg.1 NM_001097643.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200202700.hg.1 TC1200012763.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200202699.hg.1 ENST00000539585.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200202699.hg.1 NM_001097643.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200202699.hg.1 ENST00000539585.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200202699.hg.1 NM_001097643.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200202699.hg.1 TC1200012763.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200202698.hg.1 ENST00000539585.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200202698.hg.1 NM_001097643.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200202698.hg.1 ENST00000539585.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200202698.hg.1 NM_001097643.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200202698.hg.1 TC1200012763.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A047333 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A047333 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3238772 NM_001097643.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3238772 NM_001097643.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3238772 NM_001097643.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3238772 NM_001097643.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline