Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03298580_g1
          See other HSP90AB4P GE Assays ›
          Gene Symbol:
          HSP90AB4P
          Gene Name:
          heat shock protein 90 alpha family class B member 4, pseudogene
          Gene Aliases:
          HSP90Bd
          Chromosome Location:
          Chr.15: 58690473 - 58693607 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03298580_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HSP90AB4P
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AY956765.1 - 2 - 3 991 89

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream lipase C,
          hepatic type
          LIPC HDLCQ12,
          HL,
          HTGL,
          LIPH
          -122.2kb Anti-sense Chr. 15 - 58410754 - 58568952 on NCBI Build GRCh38
          Downstream uncharacterized LOC101928725 LOC101928725 75.9kb Sense Chr. 15 - 58768072 - 58770974 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons

          Panther Classification:

          Molecular Function -

          Hsp90 family chaperone

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3310718 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3933 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3932 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 50656 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3310718 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3310718 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09685885 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09685880 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30178503 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30178513 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09685882 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30178515 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30178519 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09685884 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09685886 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30178510 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16794372 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30178509 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09685881 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30178516 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16794374 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09685882 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30178519 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30178513 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09685884 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30178516 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30178515 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09685880 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09685881 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30178509 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16794372 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30178510 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16794374 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09685886 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09685885 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30178503 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09685888 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09685898 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09685894 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09685892 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09685880 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09685896 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09685882 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09685884 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09685887 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_002927_at - -
          Affymetrix Human Gene 1.0 ST Array 7989246 - -
          Affymetrix Human Gene 1.0 ST Array 7989247 - -
          Affymetrix Human Gene 1.0 ST Array 7989248 - -
          Affymetrix Human Gene 1.1 ST Array 7989245 - -
          Affymetrix Human Transcriptome Array 2.0 TC1500009576.hg.1 ENST00000631455.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009576.hg.1 ENST00000631455.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009576.hg.1 NR_073415.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009576.hg.1 NR_073415.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009576.hg.1 NR_073415.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009576.hg.1 NR_073415.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009576.hg.1 NR_073415.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009576.hg.1 ENST00000631455.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009576.hg.1 ENST00000631455.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009576.hg.1 NR_073415.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009576.hg.1 NR_073415.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009576.hg.1 NR_073415.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009576.hg.1 NR_073415.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009576.hg.1 NR_073415.2 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A052196 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A052196 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1783303 NR_002927.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1783303 AY956765.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2127098 NR_002927.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2127098 AY956765.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2127098 NR_002927.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2127098 AY956765.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1783303 NR_002927.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1783303 AY956765.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1783303 NR_002927.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1783303 AY956765.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2127098 NR_002927.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2127098 AY956765.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2127098 NR_002927.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2127098 AY956765.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1783303 NR_002927.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1783303 AY956765.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1783303 NR_002927.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1783303 AY956765.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2127098 NR_002927.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2127098 AY956765.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2127098 NR_002927.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2127098 AY956765.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1783303 NR_002927.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1783303 AY956765.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2127098 NR_002927.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2127098 AY956765.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1783303 NR_002927.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1783303 AY956765.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1783303 NR_002927.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1783303 AY956765.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2127098 NR_002927.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2127098 AY956765.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline