Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03298703_s1
          See other SNORA50C GE Assays ›
          Gene Symbol:
          SNORA50C SNHG25
          Gene Name:
          small nucleolar RNA, H/ACA box 50C
          small nucleolar RNA host gene 25
          Gene Aliases:
          ACA62, SNORA76, SNORA76C
          -
          Chromosome Location:
          Chr.17: 64146339 - 64146471 on Build GRCh38
          Chromosome Location:
          Chr.17: 64145970 - 64146309 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs03298703_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SNORA50C
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002995.1 1 63 75
          Gene Symbol: SNHG25
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA CD050382.1 - 220 75
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AI123657.1 - 1 Chr. 17 - 64146059 - 64146476 [-] on Build GRCh38 75
          GenBankmRNA CD174478.1 - 1 198 Chr. 17 - 64146223 - 64146476 [+] on Build GRCh38 75

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream small nucleolar RNA,
          C/D box 104
          SNORD104 U104 -182.0 Sense Chr. 17 - 64146078 - 64146157 on NCBI Build GRCh38
          Downstream testis expressed 2 TEX2 HT008,
          TMEM96
          962.0 Anti-sense Chr. 17 - 64147433 - 64263323 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700008593.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17000776.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4008 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10415108 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10415110 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10415112 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10415105 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10415112 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10415105 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10415108 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10415110 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10415112 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10415108 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10415089 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10415088 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10415106 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17006366 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Genome U219 Array 11763896_at - -
          Affymetrix Human Genome U219 Array 11763896_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1700008593.hg.1 ENST00000408535.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008593.hg.1 ENST00000580828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008593.hg.1 ENST00000582965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008593.hg.1 ENST00000582965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008593.hg.1 ENST00000582965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008593.hg.1 ENST00000628590.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008593.hg.1 NR_002995.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008593.hg.1 NR_132278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17010122.hg.1 ENST00000408535.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17010122.hg.1 NR_002995.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17010122.hg.1 TC17000776.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1700164299.hg.1 ENST00000408535.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700164299.hg.1 ENST00000582965.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700164299.hg.1 ENST00000628590.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700164299.hg.1 NR_002995.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700164299.hg.1 ENST00000408535.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700164299.hg.1 ENST00000582965.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700164299.hg.1 ENST00000628590.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700164299.hg.1 NR_002995.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700164299.hg.1 TC1700008593.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1700008593.hg.1 ENST00000408535.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700008593.hg.1 ENST00000580828.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008593.hg.1 ENST00000582965.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008593.hg.1 ENST00000582965.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008593.hg.1 ENST00000582965.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008593.hg.1 ENST00000628590.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008593.hg.1 NR_002995.1 Refseq
          Affymetrix Human Clariom D Assay TC1700008593.hg.1 NR_132278.1 Refseq
          Human Genome U95 Set 90086_at - -
          Human Genome U95 Set 90086_PM_at - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3237562 NR_002995.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3237562 NR_002995.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3237562 NR_002995.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3237562 NR_002995.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-gk9w8:80/100.66.76.60:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0