Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03405491_g1
          See other HCCAT5 GE Assays ›
          Gene Symbol:
          HCCAT5
          Gene Name:
          hepatocellular carcinoma associated transcript 5
          Gene Aliases:
          FJ222407, HTA
          Chromosome Location:
          Chr.16: 73092349 - 73093774 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03405491_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HCCAT5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027756.1 2 - 3 765 128
          GenBank mRNA FJ222407.1 - 2 - 3 765 128

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1600008380.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC16000609.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3418511 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_5201 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_5202 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 30568 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3418511 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3418511 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649168 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649133 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107644 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649127 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649156 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649151 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107646 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107628 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107651 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107656 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107638 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649166 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649163 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107626 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107657 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107637 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649139 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107648 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649150 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107639 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649140 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107624 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649167 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649169 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107653 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107616 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107632 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649148 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649142 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107619 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107620 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107640 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649130 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649134 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107630 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107635 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649168 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107637 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107620 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107630 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649134 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649133 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107624 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107639 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649169 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649167 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649166 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107646 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107616 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107651 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649140 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649142 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107632 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107638 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107640 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107619 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649127 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107644 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107648 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107635 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649163 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107653 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649148 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107657 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107626 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649150 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649130 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649151 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649156 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107628 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10107656 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30649139 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107671 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107610 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107632 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107697 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107648 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107669 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107651 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107635 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107617 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16912157 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107616 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107663 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107698 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107626 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107696 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107661 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107614 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107692 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107690 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107640 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107662 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107691 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107684 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107658 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107682 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107700 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107679 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16912163 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107646 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107612 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107620 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107638 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107674 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107637 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16912159 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107686 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107653 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107630 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107642 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107693 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107676 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107645 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107656 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107628 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107623 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107633 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107695 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10107665 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1570315_PM_at - -
          Affymetrix Human Genome U219 Array 11735977_PM_at - -
          Affymetrix Human Genome U219 Array 11735977_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR16008723.hg.1 NR_027756.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16008723.hg.1 TC16000609.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR16008722.hg.1 NR_027756.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16008722.hg.1 TC16000609.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR16008724.hg.1 NR_027756.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16008724.hg.1 TC16000609.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1600008380.hg.1 ENST00000569990.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008380.hg.1 ENST00000569990.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008380.hg.1 ENST00000569990.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008380.hg.1 NR_027756.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008380.hg.1 NR_027756.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008380.hg.1 NR_027756.1 Refseq
          Affymetrix Human Clariom D Assay TC1600008380.hg.1 ENST00000569990.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600008380.hg.1 ENST00000569990.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600008380.hg.1 ENST00000569990.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600008380.hg.1 NR_027756.1 Refseq
          Affymetrix Human Clariom D Assay TC1600008380.hg.1 NR_027756.1 Refseq
          Affymetrix Human Clariom D Assay TC1600008380.hg.1 NR_027756.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600163025.hg.1 ENST00000569990.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1600163025.hg.1 NR_027756.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600163025.hg.1 ENST00000569990.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1600163025.hg.1 NR_027756.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600163025.hg.1 TC1600008380.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600163027.hg.1 ENST00000569990.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1600163027.hg.1 NR_027756.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600163027.hg.1 ENST00000569990.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1600163027.hg.1 NR_027756.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600163027.hg.1 TC1600008380.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600163026.hg.1 ENST00000569990.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1600163026.hg.1 NR_027756.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600163026.hg.1 ENST00000569990.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1600163026.hg.1 NR_027756.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600163026.hg.1 TC1600008380.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 1570315_PM_at - -
          Human Genome U133 Plus 2.0 Array 1570315_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570315_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570315_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570315_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570315_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570315_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570315_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570315_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570315_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570315_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570315_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570315_at8 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A054790 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A054790 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 443480 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3242063 FJ222407.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3242063 FJ222408.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3242063 NR_027756.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3242063 BC019009.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3242063 FJ222407.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3242063 FJ222408.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3242063 NR_027756.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3242063 BC019009.2 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline