| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT HG-U133+ PM Array Plate |
1559645_PM_at |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
1559646_PM_a_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012030.hg.1 |
ENST00000429269.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012030.hg.1 |
ENST00000429269.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012030.hg.1 |
ENST00000429269.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012030.hg.1 |
ENST00000429269.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012030.hg.1 |
ENST00000429269.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012030.hg.1 |
NR_033927.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012030.hg.1 |
NR_033927.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012030.hg.1 |
NR_033927.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012030.hg.1 |
NR_033927.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012030.hg.1 |
NR_033927.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030005.hg.1 |
ENST00000429269.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030005.hg.1 |
NR_033927.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030005.hg.1 |
TC01001938.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030006.hg.1 |
ENST00000429269.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030006.hg.1 |
NR_033927.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030006.hg.1 |
TC01001938.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100188689.hg.1 |
ENST00000429269.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188689.hg.1 |
NR_033927.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188689.hg.1 |
ENST00000429269.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188689.hg.1 |
NR_033927.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188689.hg.1 |
TC0100012030.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100188692.hg.1 |
ENST00000429269.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188692.hg.1 |
NR_033927.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188692.hg.1 |
ENST00000429269.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188692.hg.1 |
NR_033927.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188692.hg.1 |
TC0100012030.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100012030.hg.1 |
ENST00000429269.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012030.hg.1 |
ENST00000429269.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012030.hg.1 |
ENST00000429269.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012030.hg.1 |
ENST00000429269.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012030.hg.1 |
ENST00000429269.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012030.hg.1 |
NR_033927.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012030.hg.1 |
NR_033927.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012030.hg.1 |
NR_033927.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012030.hg.1 |
NR_033927.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012030.hg.1 |
NR_033927.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188691.hg.1 |
ENST00000429269.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188691.hg.1 |
NR_033927.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188691.hg.1 |
ENST00000429269.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188691.hg.1 |
NR_033927.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188691.hg.1 |
TC0100012030.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100188690.hg.1 |
ENST00000429269.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188690.hg.1 |
NR_033927.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188690.hg.1 |
ENST00000429269.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188690.hg.1 |
NR_033927.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188690.hg.1 |
TC0100012030.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100188693.hg.1 |
ENST00000429269.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188693.hg.1 |
NR_033927.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188693.hg.1 |
ENST00000429269.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188693.hg.1 |
NR_033927.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188693.hg.1 |
TC0100012030.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
1559645_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1559646_PM_a_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1559646_a_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1559645_PM_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559645_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559645_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559646_a_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559645_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559646_a_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559646_a_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559645_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559646_a_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559645_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559646_a_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559645_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559645_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559646_a_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559645_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559646_a_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559645_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559646_a_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559645_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559646_a_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559645_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559646_a_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559646_a_at4 |
- |
- |