Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03644906_s1
          See other ARIH2OS GE Assays ›
          Gene Symbol:
          ARIH2OS
          Gene Name:
          ariadne homolog 2 opposite strand
          Gene Aliases:
          C3orf71
          Chromosome Location:
          Chr.3: 48917788 - 48919385 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03644906_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ARIH2OS
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001123040.1 NP_001116512.1 1 1052 80
          GenBank mRNA AK097724.1 - 1 1052 80
          DA461567.1 - 1 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300011039.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03001391.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P76018 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P76018 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 2774 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 7782 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P00700154 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P76018 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P76018 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358957 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358952 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358930 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02224492 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358959 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02224460 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358928 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358931 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358958 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358963 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358954 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358979 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358943 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02224459 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358946 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358953 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358956 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358926 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02224469 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358968 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02224486 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358974 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358932 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358973 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358969 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358980 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15435672 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02224478 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358962 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358976 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358948 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02224473 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358959 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358957 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02224486 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358956 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02224459 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358962 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358930 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358974 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358932 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358943 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358948 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358968 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358958 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358928 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358953 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358976 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02224473 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358946 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358963 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02224478 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02224460 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358952 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02224492 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358969 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358973 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02224469 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358926 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358954 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358931 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358979 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22358980 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15435672 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P76018 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P76018 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00700154 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02224488 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02224466 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02224463 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02224476 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02224479 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15435679 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02224472 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02224461 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15435672 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02224469 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02224448 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02224485 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02224452 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02224484 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02224450 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02224481 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02224465 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02224470 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02224467 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02224459 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02224491 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001123040_at - -
          Affymetrix Human Gene 1.0 ST Array 8087235 - -
          Affymetrix Human Gene 1.1 ST Array 8087234 - -
          Affymetrix HT HG-U133+ PM Array Plate 1559532_PM_at - -
          Affymetrix Human Genome U219 Array 11740715_PM_at - -
          Affymetrix Human Genome U219 Array 11740715_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1200012856.hg.1 ENST00000321694.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012856.hg.1 ENST00000321694.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012856.hg.1 ENST00000321694.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012856.hg.1 ENST00000539956.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012856.hg.1 ENST00000539956.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012856.hg.1 ENST00000539956.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012856.hg.1 ENST00000539956.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012856.hg.1 ENST00000539956.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012856.hg.1 ENST00000539956.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012856.hg.1 ENST00000621133.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012856.hg.1 ENST00000621133.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03024523.hg.1 TC03001391.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03024523.hg.1 ENST00000408959.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03024523.hg.1 NM_001123040.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011039.hg.1 ENST00000408959.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011039.hg.1 ENST00000408959.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011039.hg.1 ENST00000408959.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011039.hg.1 NM_001123040.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011039.hg.1 NM_001123040.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011039.hg.1 NM_001123040.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300181340.hg.1 ENST00000408959.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0300181340.hg.1 NM_001123040.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300181340.hg.1 ENST00000408959.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0300181340.hg.1 NM_001123040.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300181340.hg.1 TC0300011039.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0300011039.hg.1 ENST00000408959.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300011039.hg.1 ENST00000408959.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300011039.hg.1 ENST00000408959.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300011039.hg.1 NM_001123040.1 Refseq
          Affymetrix Human Clariom D Assay TC0300011039.hg.1 NM_001123040.1 Refseq
          Affymetrix Human Clariom D Assay TC0300011039.hg.1 NM_001123040.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300181341.hg.1 ENST00000408959.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0300181341.hg.1 NM_001123040.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300181341.hg.1 ENST00000408959.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0300181341.hg.1 NM_001123040.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300181341.hg.1 TC0300011039.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300181342.hg.1 ENST00000408959.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0300181342.hg.1 NM_001123040.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300181342.hg.1 ENST00000408959.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0300181342.hg.1 NM_001123040.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300181342.hg.1 TC0300011039.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1200012856.hg.1 ENST00000321694.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012856.hg.1 ENST00000321694.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012856.hg.1 ENST00000321694.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012856.hg.1 ENST00000539956.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012856.hg.1 ENST00000539956.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012856.hg.1 ENST00000539956.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012856.hg.1 ENST00000539956.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012856.hg.1 ENST00000539956.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012856.hg.1 ENST00000539956.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012856.hg.1 ENST00000621133.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012856.hg.1 ENST00000621133.1 Ensembl
          Human Genome U133 Plus 2.0 Array 1559532_at - -
          Human Genome U133 Plus 2.0 Array 1559532_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559532_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559532_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559532_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559532_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559532_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559532_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559532_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559532_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559532_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559532_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559532_at3 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A021046 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A021046 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3241670 NM_001123040.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3241670 AK097724.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3241670 NM_001123040.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3241670 AK097724.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3241670 NM_001123040.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3241670 AK097724.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3241670 NM_001123040.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3241670 AK097724.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0