| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06025034.hg.1 |
ENST00000338882.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06025034.hg.1 |
ENST00000490043.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06025034.hg.1 |
NM_001123364.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06025034.hg.1 |
TC06004135.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012829.hg.1 |
ENST00000338882.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012829.hg.1 |
ENST00000338882.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012829.hg.1 |
ENST00000338882.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012829.hg.1 |
ENST00000338882.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012829.hg.1 |
ENST00000338882.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012829.hg.1 |
ENST00000490043.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012829.hg.1 |
ENST00000490043.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012829.hg.1 |
ENST00000490043.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012829.hg.1 |
NM_001123364.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012829.hg.1 |
NM_001123364.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012829.hg.1 |
NM_001123364.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012829.hg.1 |
NM_001123364.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012829.hg.1 |
NM_001123364.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06025032.hg.1 |
ENST00000338882.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06025032.hg.1 |
NM_001123364.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06025032.hg.1 |
TC06004135.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06025037.hg.1 |
ENST00000338882.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06025037.hg.1 |
NM_001123364.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06025037.hg.1 |
TC06004135.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600188593.hg.1 |
ENST00000338882.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600188593.hg.1 |
NM_001123364.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600188593.hg.1 |
ENST00000338882.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600188593.hg.1 |
NM_001123364.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600188593.hg.1 |
TC0600012829.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600188595.hg.1 |
ENST00000338882.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600188595.hg.1 |
ENST00000490043.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600188595.hg.1 |
NM_001123364.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600188595.hg.1 |
ENST00000338882.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600188595.hg.1 |
ENST00000490043.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600188595.hg.1 |
NM_001123364.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600188595.hg.1 |
TC0600012829.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600188598.hg.1 |
ENST00000338882.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600188598.hg.1 |
NM_001123364.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600188598.hg.1 |
ENST00000338882.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600188598.hg.1 |
NM_001123364.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600188598.hg.1 |
TC0600012829.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600012829.hg.1 |
ENST00000338882.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012829.hg.1 |
ENST00000338882.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012829.hg.1 |
ENST00000338882.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012829.hg.1 |
ENST00000338882.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012829.hg.1 |
ENST00000338882.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012829.hg.1 |
ENST00000490043.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012829.hg.1 |
ENST00000490043.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012829.hg.1 |
ENST00000490043.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012829.hg.1 |
NM_001123364.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012829.hg.1 |
NM_001123364.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012829.hg.1 |
NM_001123364.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012829.hg.1 |
NM_001123364.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012829.hg.1 |
NM_001123364.2 |
Refseq |