| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008630.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008630.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008630.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008630.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008630.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008630.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008630.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008630.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008630.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008630.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008630.hg.1 |
ENST00000434409.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008630.hg.1 |
ENST00000434409.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008630.hg.1 |
ENST00000434409.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008630.hg.1 |
ENST00000434409.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008630.hg.1 |
ENST00000434409.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008630.hg.1 |
NR_024130.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008630.hg.1 |
NR_024130.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008630.hg.1 |
NR_024130.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008630.hg.1 |
NR_024130.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008630.hg.1 |
NR_024130.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008630.hg.1 |
NR_024130.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008371.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008371.hg.1 |
ENST00000434409.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008371.hg.1 |
NR_024130.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008371.hg.1 |
TC10000716.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008370.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008370.hg.1 |
ENST00000434409.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008370.hg.1 |
NR_024130.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008370.hg.1 |
TC10000716.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1000008630.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008630.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008630.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008630.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008630.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008630.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008630.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008630.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008630.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008630.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008630.hg.1 |
ENST00000434409.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008630.hg.1 |
ENST00000434409.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008630.hg.1 |
ENST00000434409.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008630.hg.1 |
ENST00000434409.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008630.hg.1 |
ENST00000434409.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008630.hg.1 |
NR_024130.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008630.hg.1 |
NR_024130.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008630.hg.1 |
NR_024130.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008630.hg.1 |
NR_024130.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008630.hg.1 |
NR_024130.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008630.hg.1 |
NR_024130.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000158886.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000158886.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000158886.hg.1 |
ENST00000434409.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000158886.hg.1 |
NR_024130.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000158886.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000158886.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000158886.hg.1 |
ENST00000434409.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000158886.hg.1 |
NR_024130.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000158886.hg.1 |
TC1000008630.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000158887.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000158887.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000158887.hg.1 |
ENST00000434409.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000158887.hg.1 |
NR_024130.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000158887.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000158887.hg.1 |
ENST00000427093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000158887.hg.1 |
ENST00000434409.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000158887.hg.1 |
NR_024130.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000158887.hg.1 |
TC1000008630.hg.1 |
Clariom D TC |