Search
Search

ZNF83| Gene Symbol: ZNF83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC017737.1 | - | 1 | 546 | 83 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC137778871 | LOC137778871 | -12.6kb | Anti-sense | Chr. 19 - 52601301 - 52635579 on NCBI Build GRCh38 | |
| Downstream | Zinc finger protein LOC122539214 | LOC122539214 | 734.0 | Sense | Chr. 19 - 52650447 - 52690496 on NCBI Build GRCh38 |
Set Membership: |
|