Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03671001_s1
          See other LOC283357 GE Assays ›
          Gene Symbol:
          LOC283357
          Gene Name:
          uncharacterized LOC283357
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03671001_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC283357
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AL833464.1 - 1 447 78

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 936334 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 802154 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 258200 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 209603 AF116637.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 209603 BC020630.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 209603 CR604926.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 209603 CR606015.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 209603 NM_018584.5 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CNV Microarray 2x400K 304726 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P11313831 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 235151_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1200009549.hg.1 ENST00000570140.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200009549.hg.1 ENST00000570140.1 Ensembl
          Human Genome U133 Plus 2.0 Array 235151_at - -
          Human Genome U133 Plus 2.0 Array 235151_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235151_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235151_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235151_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235151_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235151_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235151_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235151_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235151_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235151_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235151_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235151_at5 - -
          Human Genome U95 Set 88859_at - -
          Human Genome U95 Set 88859_PM_at - -
          Human Genome U133 Set 235151_at - -
          Human Genome U133 Set 235151_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline