| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT HG-U133+ PM Array Plate |
1563053_PM_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007171.hg.1 |
ENST00000432377.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007171.hg.1 |
ENST00000432377.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007171.hg.1 |
NR_122070.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007171.hg.1 |
NR_122070.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007171.hg.1 |
ENST00000432377.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007171.hg.1 |
ENST00000432377.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007171.hg.1 |
NR_122070.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007171.hg.1 |
NR_122070.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300150510.hg.1 |
ENST00000432377.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300150510.hg.1 |
NR_122070.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300150510.hg.1 |
ENST00000432377.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300150510.hg.1 |
NR_122070.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300150510.hg.1 |
TC0300007171.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300150509.hg.1 |
ENST00000432377.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300150509.hg.1 |
NR_122070.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300150509.hg.1 |
ENST00000432377.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300150509.hg.1 |
NR_122070.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300150509.hg.1 |
TC0300007171.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
1563053_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1563053_PM_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1563053_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1563053_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1563053_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1563053_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1563053_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1563053_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1563053_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1563053_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1563053_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1563053_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1563053_at10 |
- |
- |