Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03678783_m1
          See other FLVCR1-AS1 GE Assays ›
          Gene Symbol:
          FLVCR1-AS1
          Gene Name:
          FLVCR1 antisense RNA 1 (head to head)
          Gene Aliases:
          LQK1, NCRNA00292
          Chromosome Location:
          Chr.1: 212856604 - 212858138 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03678783_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FLVCR1-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027286.1 1 - 2 370 79
          GenBank mRNA AY030239.1 - 1 - 2 309 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100017262.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01003812.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Homo sapiens Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3299811 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P190404 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 15669 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 29509 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3299811 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3299811 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15172085 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00780232 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00780230 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00780229 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20843338 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00780227 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20843338 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00780232 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00780229 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00780230 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00780227 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15172085 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P190404 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P190404 - -
          Agilent Human 1B Microarray G4111A A_32_P190408 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00780205 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00780232 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00780202 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00780204 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15172085 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00780225 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00780227 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00780229 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00780207 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7924188 - -
          Affymetrix Human Gene 1.0 ST Array 7924189 - -
          Affymetrix Human Gene 1.0 ST Array 7924187 - -
          Affymetrix Human Gene 1.0 ST Array 7924186 - -
          Affymetrix Human Gene 1.1 ST Array 7924184 - -
          Affymetrix HT HG-U133+ PM Array Plate 235126_PM_at - -
          Affymetrix Human Genome U219 Array 11758579_s_at - -
          Affymetrix Human Genome U219 Array 11744731_PM_at - -
          Affymetrix Human Genome U219 Array 11744731_at - -
          Affymetrix Human Genome U219 Array 11758579_PM_s_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR01058234.hg.1 TC01003812.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01058234.hg.1 NR_027285.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01058234.hg.1 NR_027286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01058237.hg.1 TC01003812.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01058237.hg.1 NR_027285.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01058237.hg.1 NR_027286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01058236.hg.1 TC01003812.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01058236.hg.1 ENST00000356684.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01058236.hg.1 NR_027285.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01058236.hg.1 NR_027286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 ENST00000356684.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 ENST00000356684.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 ENST00000356684.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 ENST00000356684.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 ENST00000424044.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 ENST00000424044.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 NR_027285.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 NR_027285.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 NR_027285.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 NR_027285.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 NR_027286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 NR_027286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 NR_027286.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 ENST00000356684.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 ENST00000356684.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 ENST00000356684.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 ENST00000356684.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 ENST00000424044.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 ENST00000424044.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 NR_027285.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 NR_027285.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 NR_027285.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 NR_027285.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 NR_027286.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 NR_027286.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 NR_027286.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100230667.hg.1 ENST00000424044.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100230667.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100230667.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100230667.hg.1 NR_027285.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100230667.hg.1 NR_027286.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100230667.hg.1 ENST00000424044.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100230667.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100230667.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100230667.hg.1 NR_027285.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100230667.hg.1 NR_027286.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100230667.hg.1 TC0100017262.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100230673.hg.1 NR_027285.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100230673.hg.1 NR_027286.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100230673.hg.1 NR_027285.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100230673.hg.1 NR_027286.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100230673.hg.1 TC0100017262.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100230670.hg.1 ENST00000356684.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100230670.hg.1 ENST00000356684.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100230670.hg.1 ENST00000424044.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100230670.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100230670.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100230670.hg.1 NR_027285.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100230670.hg.1 NR_027286.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100230670.hg.1 ENST00000356684.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100230670.hg.1 ENST00000356684.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100230670.hg.1 ENST00000424044.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100230670.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100230670.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100230670.hg.1 NR_027285.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100230670.hg.1 NR_027286.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100230670.hg.1 TC0100017262.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 235126_PM_at - -
          Human Genome U133 Plus 2.0 Array 235126_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235126_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235126_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235126_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235126_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235126_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235126_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235126_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235126_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235126_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235126_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235126_at11 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A007495 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A010932 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A010932 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A007495 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1750368 NR_027285.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1750368 NR_027286.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1750368 AY030238.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1750368 AY030239.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1683183 - -
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1668771 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1750368 NR_027285.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1750368 NR_027286.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1750368 AY030238.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1750368 AY030239.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1683183 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1668771 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1782178 NR_027285.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1782178 AY030238.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1668771 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1750368 NR_027285.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1750368 NR_027286.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1750368 AY030238.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1750368 AY030239.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1782178 NR_027285.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1782178 AY030238.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1668771 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1750368 NR_027285.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1750368 NR_027286.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1750368 AY030238.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1750368 AY030239.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline