| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008848.hg.1 |
ENST00000566370.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008848.hg.1 |
NR_047493.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008848.hg.1 |
ENST00000566370.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008848.hg.1 |
NR_047493.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300159699.hg.1 |
ENST00000566370.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300159699.hg.1 |
NR_047493.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300159699.hg.1 |
ENST00000566370.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300159699.hg.1 |
NR_047493.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300159699.hg.1 |
TC1300008848.hg.1 |
Clariom D TC |