Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03805942_s1
          See other PPP1R3G GE Assays ›
          Gene Symbol:
          PPP1R3G
          Gene Name:
          protein phosphatase 1 regulatory subunit 3G
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 5085486 - 5087221 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03805942_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PPP1R3G
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145115.2 NP_001138587.1 1 451 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600006805.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000047.hg.1

          Set Membership:

          Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          phosphatase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          glucose homeostasis
          positive regulation of glycogen biosynthetic process
          positive regulation of glycogen (starch) synthase activity
          protein phosphatase binding
          glycogen binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 433217 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 207151 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3353996 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1997 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1996 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 42931 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P37501049 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3353996 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3353996 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710862 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523888 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523909 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523902 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710842 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710822 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523872 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523871 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710874 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710836 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523873 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523891 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523915 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710825 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523900 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710831 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523892 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523882 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523906 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523895 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710833 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710873 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710823 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710827 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710817 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710846 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523898 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523901 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710860 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523903 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523908 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710841 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523905 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710827 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710874 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523882 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523891 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710860 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523900 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523915 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710817 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710836 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523873 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710842 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710822 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523903 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523872 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710862 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710841 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710846 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523888 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523901 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710825 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523892 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710823 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523905 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710873 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710833 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523871 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523909 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523895 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523906 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523908 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523902 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24710831 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04523898 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523949 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523920 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523909 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523903 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523915 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523946 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523969 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523884 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523871 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523964 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15780062 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523890 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523886 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523955 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523899 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523892 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523931 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523895 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523880 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523878 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15780049 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523952 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523904 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523926 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523873 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523967 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15780067 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523919 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523869 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523882 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523913 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523918 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523957 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523928 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15780051 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523877 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523938 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523923 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523897 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523933 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523972 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523961 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523942 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523901 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523911 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523902 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523893 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523944 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04523906 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Genome U219 Array 11735684_at - -
          Affymetrix Human Genome U219 Array 11735684_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0600006805.hg.1 ENST00000405617.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006805.hg.1 ENST00000405617.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006805.hg.1 ENST00000405617.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006805.hg.1 ENST00000405617.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006805.hg.1 NM_001145115.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006805.hg.1 NM_001145115.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06000424.hg.1 TC06000047.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06000424.hg.1 ENST00000405617.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000424.hg.1 NM_001145115.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600146719.hg.1 ENST00000405617.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146719.hg.1 NM_001145115.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600146719.hg.1 ENST00000405617.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146719.hg.1 NM_001145115.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600146719.hg.1 TC0600006805.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0600006805.hg.1 ENST00000405617.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600006805.hg.1 ENST00000405617.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600006805.hg.1 ENST00000405617.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600006805.hg.1 ENST00000405617.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600006805.hg.1 NM_001145115.2 Refseq
          Affymetrix Human Clariom D Assay TC0600006805.hg.1 NM_001145115.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600146718.hg.1 ENST00000405617.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146718.hg.1 NM_001145115.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600146718.hg.1 ENST00000405617.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146718.hg.1 NM_001145115.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600146718.hg.1 TC0600006805.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A030933 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A030933 - -
          Array Name Probe IDs Targets Target Type
          Illumina human-6 v2.0 expression beadchip ILMN_1800716 NM_001145115.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1800716 NM_001145115.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline