Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03806111_m1
          See other TRIM77 GE Assays ›
          Gene Symbol:
          TRIM77
          Gene Name:
          tripartite motif containing 77
          Gene Aliases:
          TRIM77P
          Chromosome Location:
          Chr.11: 89710299 - 89717872 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03806111_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TRIM77
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001146162.1 NP_001139634.1 5 - 6 855 103
          NM_001271942.1 NP_001258871.1 4 - 5 624 103
          GenBank mRNA JF968446.1 - 4 - 5 273 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100008713.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11000889.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 Homo sapiens 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Process(es)

          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human CGH Microarray 4x180K 3752 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 224712 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 191003 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 407282 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 1534 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3235048 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_2106 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_2105 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 25552 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P11180430 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3235048 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3235048 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28360668 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07937259 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28360646 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16486418 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28360638 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07937260 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28360653 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28360626 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28360668 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07937260 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07937259 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16486418 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28360653 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28360646 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28360638 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28360626 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07937271 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07937263 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07937258 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07937276 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P11180430 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Genome U219 Array 11742492_PM_at - -
          Affymetrix Human Genome U219 Array 11742492_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 ENST00000398290.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 ENST00000398290.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 ENST00000398290.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 ENST00000398290.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 ENST00000398290.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 ENST00000398290.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 ENST00000398290.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 ENST00000398290.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 ENST00000398290.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 ENST00000534392.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 ENST00000534392.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 ENST00000534392.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 ENST00000534392.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 ENST00000534392.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 ENST00000534392.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 NM_001146162.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 NM_001146162.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 NM_001146162.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 NM_001146162.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 NM_001146162.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 NM_001146162.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 NM_001146162.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 NM_001146162.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 NM_001146162.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 NM_001271942.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 NM_001271942.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 NM_001271942.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 NM_001271942.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 NM_001271942.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 NM_001271942.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 NM_001271942.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008713.hg.1 NM_001271942.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11011188.hg.1 NM_001146162.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11011188.hg.1 TC11000889.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100010790.hg.1 ENST00000524985.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010790.hg.1 ENST00000524985.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010790.hg.1 ENST00000524985.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010790.hg.1 ENST00000524985.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100164399.hg.1 ENST00000398290.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100164399.hg.1 ENST00000534392.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100164399.hg.1 NM_001146162.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100164399.hg.1 NM_001271942.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100164399.hg.1 ENST00000398290.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100164399.hg.1 ENST00000534392.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100164399.hg.1 NM_001146162.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100164399.hg.1 NM_001271942.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100164399.hg.1 TC1100008713.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 ENST00000398290.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 ENST00000398290.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 ENST00000398290.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 ENST00000398290.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 ENST00000398290.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 ENST00000398290.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 ENST00000398290.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 ENST00000398290.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 ENST00000398290.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 ENST00000534392.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 ENST00000534392.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 ENST00000534392.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 ENST00000534392.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 ENST00000534392.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 ENST00000534392.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 NM_001146162.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 NM_001146162.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 NM_001146162.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 NM_001146162.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 NM_001146162.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 NM_001146162.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 NM_001146162.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 NM_001146162.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 NM_001146162.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 NM_001271942.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 NM_001271942.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 NM_001271942.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 NM_001271942.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 NM_001271942.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 NM_001271942.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 NM_001271942.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008713.hg.1 NM_001271942.1 Refseq
          Affymetrix Human Clariom D Assay TC1100010790.hg.1 ENST00000524985.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010790.hg.1 ENST00000524985.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010790.hg.1 ENST00000524985.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010790.hg.1 ENST00000524985.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100164400.hg.1 ENST00000398290.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100164400.hg.1 ENST00000534392.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100164400.hg.1 NM_001146162.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100164400.hg.1 NM_001271942.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100164400.hg.1 ENST00000398290.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100164400.hg.1 ENST00000534392.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100164400.hg.1 NM_001146162.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100164400.hg.1 NM_001271942.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100164400.hg.1 TC1100008713.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100164401.hg.1 ENST00000398290.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100164401.hg.1 NM_001146162.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100164401.hg.1 NM_001271942.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100164401.hg.1 ENST00000398290.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100164401.hg.1 NM_001146162.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100164401.hg.1 NM_001271942.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100164401.hg.1 TC1100008713.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046841 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046841 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 171595 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1771171 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1771171 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1732096 NM_001146162.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1732096 NM_001146162.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline