Search
Search

NONOP2| Gene Symbol: NONOP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK124741.1 | - | 1 | 3237 | 76 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
REL proto-oncogene, NF-kB subunit |
REL |
C-Rel, HIVEN86A, IMD92 |
-4.0kb | Sense | Chr. 2 - 60881574 - 60931612 on NCBI Build GRCh38 |
| Downstream | pseudouridine synthase 10 | PUS10 |
CCDC139, DOBI, Hup10 |
4.0kb | Anti-sense | Chr. 2 - 60940223 - 61018259 on NCBI Build GRCh38 |
Set Membership: |
|