Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03917769_s1
          See other LINC00421 GE Assays ›
          Gene Symbol:
          LINC00421
          Gene Name:
          long intergenic non-protein coding RNA 421
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 19345049 - 19346749 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03917769_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00421
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_034026.1 2 250 107
          GenBank mRNA BC022083.2 - 2 250 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1300006468.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29148423 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29148419 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29148460 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29148461 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29148442 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08705553 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16612879 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08705563 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29148445 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08705556 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29148410 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29148414 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08705561 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16612892 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29148418 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29148424 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08705555 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29148452 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29148435 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P920268 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 35854 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P920268 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P920268 - -
          Agilent Human 1B Microarray G4111A A_32_P920268 - -
          Agilent Human 1B Microarray G4111A A_32_P506665 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1555196_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1300006468.hg.1 ENST00000413833.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006468.hg.1 ENST00000413833.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006468.hg.1 ENST00000413833.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006468.hg.1 ENST00000413833.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006468.hg.1 ENST00000413833.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006468.hg.1 NR_034026.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006468.hg.1 NR_034026.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006468.hg.1 NR_034026.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006468.hg.1 NR_034026.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006468.hg.1 NR_034026.1 Refseq
          Affymetrix Human Clariom D Assay TC1300006468.hg.1 ENST00000413833.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300006468.hg.1 ENST00000413833.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300006468.hg.1 ENST00000413833.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300006468.hg.1 ENST00000413833.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300006468.hg.1 ENST00000413833.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300006468.hg.1 NR_034026.1 Refseq
          Affymetrix Human Clariom D Assay TC1300006468.hg.1 NR_034026.1 Refseq
          Affymetrix Human Clariom D Assay TC1300006468.hg.1 NR_034026.1 Refseq
          Affymetrix Human Clariom D Assay TC1300006468.hg.1 NR_034026.1 Refseq
          Affymetrix Human Clariom D Assay TC1300006468.hg.1 NR_034026.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300144180.hg.1 ENST00000413833.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300144180.hg.1 NR_034026.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300144180.hg.1 ENST00000413833.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300144180.hg.1 NR_034026.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300144180.hg.1 TC1300006468.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300144182.hg.1 ENST00000413833.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300144182.hg.1 NR_034026.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300144182.hg.1 ENST00000413833.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300144182.hg.1 NR_034026.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300144182.hg.1 TC1300006468.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300144181.hg.1 ENST00000413833.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300144181.hg.1 NR_034026.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300144181.hg.1 ENST00000413833.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300144181.hg.1 NR_034026.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300144181.hg.1 TC1300006468.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 1555196_PM_at - -
          Human Genome U133 Plus 2.0 Array 1555196_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555196_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555196_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555196_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555196_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555196_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555196_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555196_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555196_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555196_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555196_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555196_at1 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A049898 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A049898 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 223810 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline