| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT HG-U133+ PM Array Plate |
1557133_PM_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11734170_PM_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11734170_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008853.hg.1 |
ENST00000370535.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008853.hg.1 |
NR_028344.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008853.hg.1 |
NR_028345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008853.hg.1 |
TC0X000683.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008854.hg.1 |
ENST00000370535.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008854.hg.1 |
ENST00000498732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008854.hg.1 |
NR_028344.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008854.hg.1 |
NR_028345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008854.hg.1 |
TC0X000683.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
ENST00000370535.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
ENST00000370535.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
ENST00000370535.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
ENST00000498732.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
ENST00000498732.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
ENST00000498732.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
ENST00000498732.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
ENST00000498732.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
ENST00000498732.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
ENST00000602535.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
ENST00000602535.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
ENST00000602535.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
ENST00000602535.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
ENST00000602830.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
ENST00000602830.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
ENST00000602830.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
ENST00000607004.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
ENST00000607004.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
ENST00000607004.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
NR_028344.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
NR_028344.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
NR_028344.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
NR_028345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
NR_028345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
NR_028345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
NR_028345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
NR_104228.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
NR_104228.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
NR_104228.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
NR_104228.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
NR_104228.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008593.hg.1 |
NR_104228.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008855.hg.1 |
ENST00000370535.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008855.hg.1 |
ENST00000498732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008855.hg.1 |
NR_028344.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008855.hg.1 |
NR_028345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008855.hg.1 |
TC0X000683.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008852.hg.1 |
NR_028344.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008852.hg.1 |
NR_028345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008852.hg.1 |
TC0X000683.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008856.hg.1 |
NR_028345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008856.hg.1 |
TC0X000683.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0X00158332.hg.1 |
ENST00000370535.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158332.hg.1 |
ENST00000498732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158332.hg.1 |
ENST00000607004.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158332.hg.1 |
NR_028344.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158332.hg.1 |
NR_028345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158332.hg.1 |
NR_104228.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158332.hg.1 |
ENST00000370535.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158332.hg.1 |
ENST00000498732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158332.hg.1 |
ENST00000607004.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158332.hg.1 |
NR_028344.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158332.hg.1 |
NR_028345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158332.hg.1 |
NR_104228.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158332.hg.1 |
TC0X00008593.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
ENST00000370535.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
ENST00000370535.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
ENST00000370535.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
ENST00000498732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
ENST00000498732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
ENST00000498732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
ENST00000498732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
ENST00000498732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
ENST00000498732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
ENST00000602535.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
ENST00000602535.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
ENST00000602535.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
ENST00000602535.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
ENST00000602830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
ENST00000602830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
ENST00000602830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
ENST00000607004.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
ENST00000607004.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
ENST00000607004.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
NR_028344.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
NR_028344.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
NR_028344.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
NR_028345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
NR_028345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
NR_028345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
NR_028345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
NR_104228.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
NR_104228.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
NR_104228.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
NR_104228.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
NR_104228.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008593.hg.1 |
NR_104228.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158333.hg.1 |
ENST00000607004.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158333.hg.1 |
NR_028345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158333.hg.1 |
ENST00000607004.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158333.hg.1 |
NR_028345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158333.hg.1 |
TC0X00008593.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00158334.hg.1 |
NR_028345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158334.hg.1 |
NR_028345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158334.hg.1 |
TC0X00008593.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00158331.hg.1 |
ENST00000370535.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158331.hg.1 |
ENST00000498732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158331.hg.1 |
ENST00000607004.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158331.hg.1 |
NR_028344.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158331.hg.1 |
NR_028345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158331.hg.1 |
NR_104228.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158331.hg.1 |
ENST00000370535.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158331.hg.1 |
ENST00000498732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158331.hg.1 |
ENST00000607004.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158331.hg.1 |
NR_028344.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158331.hg.1 |
NR_028345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158331.hg.1 |
NR_104228.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158331.hg.1 |
TC0X00008593.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
1557133_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1557133_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557133_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557133_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557133_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557133_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557133_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557133_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557133_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557133_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557133_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557133_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557133_at5 |
- |
- |