| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11005367.hg.1 |
ENST00000356737.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11005367.hg.1 |
ENST00000538490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11005367.hg.1 |
NM_001164379.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11005367.hg.1 |
TC11000410.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11005368.hg.1 |
ENST00000356737.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11005368.hg.1 |
ENST00000538490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11005368.hg.1 |
NM_001164379.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11005368.hg.1 |
TC11000410.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007490.hg.1 |
ENST00000538490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007490.hg.1 |
ENST00000538490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007490.hg.1 |
ENST00000538490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007490.hg.1 |
NM_001164379.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007490.hg.1 |
NM_001164379.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007490.hg.1 |
NM_001164379.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100153414.hg.1 |
ENST00000538490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100153414.hg.1 |
NM_001164379.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100153414.hg.1 |
ENST00000538490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100153414.hg.1 |
NM_001164379.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100153414.hg.1 |
TC1100007490.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100007490.hg.1 |
ENST00000538490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007490.hg.1 |
ENST00000538490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007490.hg.1 |
ENST00000538490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007490.hg.1 |
NM_001164379.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007490.hg.1 |
NM_001164379.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007490.hg.1 |
NM_001164379.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100153415.hg.1 |
ENST00000538490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100153415.hg.1 |
NM_001164379.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100153415.hg.1 |
ENST00000538490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100153415.hg.1 |
NM_001164379.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100153415.hg.1 |
TC1100007490.hg.1 |
Clariom D TC |