| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
ENST00000401731.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
ENST00000401731.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
ENST00000401731.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
ENST00000401731.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
ENST00000401731.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
ENST00000401731.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
ENST00000401731.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
ENST00000599715.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
ENST00000599715.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
ENST00000599715.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
ENST00000602225.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
ENST00000602225.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
ENST00000602225.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
NM_001291485.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
NM_001291485.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
NM_001291485.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
NM_001291485.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
NM_001291485.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
NM_001291485.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
NM_001291485.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
NM_001291485.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010765.hg.1 |
NM_001291485.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021222.hg.1 |
ENST00000006724.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021222.hg.1 |
ENST00000401731.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021222.hg.1 |
ENST00000412062.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021222.hg.1 |
NM_006890.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021222.hg.1 |
TC19001560.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021223.hg.1 |
ENST00000006724.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021223.hg.1 |
ENST00000338196.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021223.hg.1 |
ENST00000401731.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021223.hg.1 |
ENST00000412062.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021223.hg.1 |
NM_006890.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021223.hg.1 |
TC19001560.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021224.hg.1 |
ENST00000006724.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021224.hg.1 |
ENST00000338196.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021224.hg.1 |
ENST00000401731.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021224.hg.1 |
NM_006890.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021224.hg.1 |
TC19001560.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021225.hg.1 |
ENST00000006724.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021225.hg.1 |
ENST00000338196.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021225.hg.1 |
ENST00000401731.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021225.hg.1 |
ENST00000412062.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021225.hg.1 |
NM_006890.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021225.hg.1 |
TC19001560.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021226.hg.1 |
ENST00000006724.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021226.hg.1 |
ENST00000338196.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021226.hg.1 |
ENST00000401731.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021226.hg.1 |
ENST00000412062.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021226.hg.1 |
NM_006890.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021226.hg.1 |
TC19001560.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
ENST00000401731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
ENST00000401731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
ENST00000401731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
ENST00000401731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
ENST00000401731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
ENST00000401731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
ENST00000401731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
ENST00000599715.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
ENST00000599715.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
ENST00000599715.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
ENST00000602225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
ENST00000602225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
ENST00000602225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
NM_001291485.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
NM_001291485.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
NM_001291485.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
NM_001291485.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
NM_001291485.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
NM_001291485.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
NM_001291485.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
NM_001291485.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010765.hg.1 |
NM_001291485.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900189003.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189003.hg.1 |
ENST00000401731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189003.hg.1 |
ENST00000602225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189003.hg.1 |
NM_001291485.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900189003.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189003.hg.1 |
ENST00000401731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189003.hg.1 |
ENST00000602225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189003.hg.1 |
NM_001291485.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900189003.hg.1 |
TC1900010765.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900189006.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189006.hg.1 |
ENST00000401731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189006.hg.1 |
NM_001291485.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900189006.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189006.hg.1 |
ENST00000401731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189006.hg.1 |
NM_001291485.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900189006.hg.1 |
TC1900010765.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900189000.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189000.hg.1 |
ENST00000401731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189000.hg.1 |
NM_001291485.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900189000.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189000.hg.1 |
ENST00000401731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189000.hg.1 |
NM_001291485.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900189000.hg.1 |
TC1900010765.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900189002.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189002.hg.1 |
ENST00000401731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189002.hg.1 |
ENST00000599715.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189002.hg.1 |
ENST00000602225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189002.hg.1 |
NM_001291485.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900189002.hg.1 |
ENST00000006724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189002.hg.1 |
ENST00000401731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189002.hg.1 |
ENST00000599715.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189002.hg.1 |
ENST00000602225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189002.hg.1 |
NM_001291485.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900189002.hg.1 |
TC1900010765.hg.1 |
Clariom D TC |