Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03989295_s1
          See other POM121L2 GE Assays ›
          Gene Symbol:
          POM121L2
          Gene Name:
          POM121 transmembrane nucleoporin like 2
          Gene Aliases:
          POM121-L, POM121L
          Chromosome Location:
          Chr.6: 27309063 - 27312232 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03989295_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: POM121L2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_033482.3 NP_258443.2 1 3056 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600011192.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06001396.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene POM121L2

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          RNA export from nucleus
          protein import into nucleus
          nucleocytoplasmic transporter activity
          nuclear localization sequence binding
          structural constituent of nuclear pore

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 841151 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3308476 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_2976 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_2977 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 702 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3308476 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3308476 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628236 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628239 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628245 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628242 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628248 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628237 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628226 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628250 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628235 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628228 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628243 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24818278 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628233 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24818295 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628247 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628246 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24818295 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628242 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628245 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628236 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24818278 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628243 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628233 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628228 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628250 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628247 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628237 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628226 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628239 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628235 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628248 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04628246 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P01446479 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15796185 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04628142 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04628249 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04628243 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04628244 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04628141 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04628246 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15796188 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15796176 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15796186 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04628247 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15796187 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04628143 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04628245 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15796184 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04628248 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P01446479 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628275 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628268 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628246 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628201 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628212 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628245 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628224 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628206 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628198 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628250 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15796179 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628243 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628257 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628191 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628237 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628260 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628219 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628204 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628271 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628226 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628233 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628255 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628259 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628248 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628266 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628209 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628196 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628239 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628221 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628235 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628214 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628264 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628242 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628207 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628229 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04628251 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P01446479 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.1 ST Array 8124495 - -
          Affymetrix HT HG-U133+ PM Array Plate 216582_PM_at - -
          Affymetrix Human Genome U219 Array 11758549_s_at - -
          Affymetrix Human Genome U219 Array 11757091_PM_at - -
          Affymetrix Human Genome U219 Array 11758549_PM_s_at - -
          Affymetrix Human Genome U219 Array 11757091_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR06016283.hg.1 TC06001396.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06016283.hg.1 ENST00000377451.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06016283.hg.1 ENST00000444565.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06016283.hg.1 NM_033482.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011192.hg.1 ENST00000429945.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011192.hg.1 ENST00000429945.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011192.hg.1 ENST00000444565.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011192.hg.1 ENST00000444565.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011192.hg.1 ENST00000444565.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011192.hg.1 NM_033482.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011192.hg.1 NM_033482.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011192.hg.1 NM_033482.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011192.hg.1 NM_033482.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06016285.hg.1 TC06001396.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06016285.hg.1 NM_033482.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06016284.hg.1 TC06001396.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06016284.hg.1 ENST00000377451.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06016284.hg.1 ENST00000444565.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06016284.hg.1 NM_033482.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06016281.hg.1 TC06001396.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06016281.hg.1 ENST00000377451.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06016281.hg.1 ENST00000444565.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06016281.hg.1 NM_033482.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06016282.hg.1 TC06001396.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06016282.hg.1 ENST00000444565.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06016282.hg.1 NM_033482.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600175596.hg.1 ENST00000444565.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600175596.hg.1 NM_033482.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600175596.hg.1 ENST00000444565.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600175596.hg.1 NM_033482.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600175596.hg.1 TC0600011192.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600175599.hg.1 NM_033482.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600175599.hg.1 NM_033482.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600175599.hg.1 TC0600011192.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600175597.hg.1 ENST00000429945.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600175597.hg.1 ENST00000444565.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600175597.hg.1 NM_033482.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600175597.hg.1 ENST00000429945.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600175597.hg.1 ENST00000444565.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600175597.hg.1 NM_033482.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600175597.hg.1 TC0600011192.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600175598.hg.1 ENST00000444565.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600175598.hg.1 NM_033482.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600175598.hg.1 ENST00000444565.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600175598.hg.1 NM_033482.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600175598.hg.1 TC0600011192.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0600011192.hg.1 ENST00000429945.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600011192.hg.1 ENST00000429945.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600011192.hg.1 ENST00000444565.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600011192.hg.1 ENST00000444565.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600011192.hg.1 ENST00000444565.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600011192.hg.1 NM_033482.3 Refseq
          Affymetrix Human Clariom D Assay TC0600011192.hg.1 NM_033482.3 Refseq
          Affymetrix Human Clariom D Assay TC0600011192.hg.1 NM_033482.3 Refseq
          Affymetrix Human Clariom D Assay TC0600011192.hg.1 NM_033482.3 Refseq
          Human Genome U133 Plus 2.0 Array 216582_PM_at - -
          Human Genome U133 Plus 2.0 Array 216582_at - -
          Human Genome U133A 2.0 Array 216582_at - -
          Human Genome U133A 2.0 Array 216582_PM_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_033482_at - -
          Human Genome U95 Set 31747_g_at - -
          Human Genome U95 Set 31746_at - -
          Human Genome U95 Set 31747_PM_g_at - -
          Human Genome U95 Set 31748_at - -
          Human Genome U95 Set 31746_PM_at - -
          Human Genome U95 Set 31748_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216582_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216582_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216582_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216582_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216582_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216582_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216582_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216582_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216582_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216582_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216582_at4 - -
          Human Genome U133 Set 216582_PM_at - -
          Human Genome U133 Set 216582_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A029556 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A029556 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3304257 NM_033482.3 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline