| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11008368.hg.1 |
ENST00000327259.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11008368.hg.1 |
NM_153266.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11008368.hg.1 |
TC11000659.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11008365.hg.1 |
ENST00000327259.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11008365.hg.1 |
NM_153266.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11008365.hg.1 |
TC11000659.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11008367.hg.1 |
ENST00000327259.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11008367.hg.1 |
NM_153266.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11008367.hg.1 |
TC11000659.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008071.hg.1 |
ENST00000327259.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008071.hg.1 |
ENST00000327259.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008071.hg.1 |
ENST00000327259.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008071.hg.1 |
ENST00000327259.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008071.hg.1 |
ENST00000327259.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008071.hg.1 |
NM_153266.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008071.hg.1 |
NM_153266.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008071.hg.1 |
NM_153266.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008071.hg.1 |
NM_153266.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008071.hg.1 |
ENST00000327259.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008071.hg.1 |
ENST00000327259.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008071.hg.1 |
ENST00000327259.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008071.hg.1 |
ENST00000327259.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008071.hg.1 |
ENST00000327259.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008071.hg.1 |
NM_153266.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008071.hg.1 |
NM_153266.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008071.hg.1 |
NM_153266.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008071.hg.1 |
NM_153266.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100158918.hg.1 |
ENST00000327259.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100158918.hg.1 |
NM_153266.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100158918.hg.1 |
ENST00000327259.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100158918.hg.1 |
NM_153266.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100158918.hg.1 |
TC1100008071.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100158919.hg.1 |
ENST00000327259.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100158919.hg.1 |
NM_153266.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100158919.hg.1 |
ENST00000327259.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100158919.hg.1 |
NM_153266.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100158919.hg.1 |
TC1100008071.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100158920.hg.1 |
ENST00000327259.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100158920.hg.1 |
NM_153266.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100158920.hg.1 |
ENST00000327259.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100158920.hg.1 |
NM_153266.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100158920.hg.1 |
TC1100008071.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100158917.hg.1 |
ENST00000327259.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100158917.hg.1 |
ENST00000327259.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100158917.hg.1 |
TC1100008071.hg.1 |
Clariom D TC |