Search
Search

CSHL1| Gene Symbol: CSHL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318.4 | NP_001309.3 | 3 - 4 | 359 | 73 | |
| NM_001321067.2 | NP_001307996.1 | 3 - 4 | 355 | 73 | ||
| NM_001321068.2 | NP_001307997.1 | 4 - 5 | 349 | 73 | ||
| NM_001321069.2 | NP_001307998.1 | 4 - 5 | 418 | 73 | ||
| NM_022579.3 | NP_072101.1 | 4 - 5 | 535 | 73 | ||
| NM_022580.3 | NP_072102.1 | 3 - 4 | 286 | 73 | ||
| NM_022581.3 | NP_072103.1 | 4 - 5 | 466 | 73 | ||
| GenBank mRNA | AX772828.1 | - | 4 - 5 | 433 | 73 | |
| BC029365.1 | - | 3 - 4 | 359 | 73 | ||
| BC119747.2 | - | 3 - 4 | 356 | 73 | ||
| BU752103.1 | - | 1 - 2 | 327 | 73 | ||
| BX426214.2 | - | 4 - 5 | 401 | 73 | ||
| DR004701.1 | - | 4 - 5 | 329 | 73 | ||
| GQ891442.1 | - | 4 - 5 | 466 | 73 | ||
| LM644139.1 | - | 3 - 4 | 458 | 73 | ||
Set Membership: |
|