Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04187239_m1
          See other ID2 GE Assays ›
          Gene Symbol:
          ID2
          Gene Name:
          inhibitor of DNA binding 2, HLH protein
          Gene Aliases:
          GIG8, ID2A, ID2H, bHLHb26
          Chromosome Location:
          Chr.2: 8681983 - 8684453 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04187239_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ID2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002166.4 NP_002157.2 2 - 3 595 81
          GenBank mRNA AK222682.1 - 2 - 3 522 81
          BC030639.2 - 2 - 3 522 4820416 81
          D13891.1 - 2 - 3 508 81
          M97796.1 - 2 - 3 520 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200006627.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02000041.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ID2

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          metanephros development
          natural killer cell differentiation
          thigmotaxis
          membranous septum morphogenesis
          bundle of His development
          transcription, DNA-templated
          multicellular organism development
          adult locomotory behavior
          positive regulation of gene expression
          negative regulation of gene expression
          oligodendrocyte development
          regulation of lipid metabolic process
          olfactory bulb development
          circadian regulation of gene expression
          mammary gland epithelial cell proliferation
          regulation of circadian rhythm
          entrainment of circadian clock by photoperiod
          enucleate erythrocyte differentiation
          negative regulation of DNA binding
          negative regulation of sequence-specific DNA binding transcription factor activity
          locomotor rhythm
          negative regulation of B cell differentiation
          positive regulation of fat cell differentiation
          positive regulation of erythrocyte differentiation
          positive regulation of macrophage differentiation
          negative regulation of neuron differentiation
          negative regulation of osteoblast differentiation
          positive regulation of blood pressure
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          cell maturation
          Peyer's patch development
          embryonic digestive tract morphogenesis
          positive regulation of smooth muscle cell proliferation
          neuron fate commitment
          cell morphogenesis involved in neuron differentiation
          positive regulation of astrocyte differentiation
          negative regulation of oligodendrocyte differentiation
          adipose tissue development
          mammary gland alveolus development
          epithelial cell differentiation involved in mammary gland alveolus development
          endodermal digestive tract morphogenesis
          positive regulation of cell cycle arrest
          cellular response to lithium ion
          positive regulation of transcription involved in G1/S transition of mitotic cell cycle
          cellular senescence
          regulation of G1/S transition of mitotic cell cycle
          negative regulation of neural precursor cell proliferation
          protein binding
          ion channel binding
          protein dimerization activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02000479.hg.1 TC02000041.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02000479.hg.1 ENST00000234091.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02000479.hg.1 ENST00000331129.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02000479.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02000479.hg.1 NM_002166.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02000481.hg.1 TC02000041.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02000481.hg.1 ENST00000234091.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02000481.hg.1 ENST00000331129.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02000481.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02000481.hg.1 NM_002166.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 ENST00000331129.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 ENST00000331129.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 ENST00000331129.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 ENST00000331129.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 ENST00000472142.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 ENST00000472142.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 ENST00000472142.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 NM_002166.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 NM_002166.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 NM_002166.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 NM_002166.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 NM_002166.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200006627.hg.1 NM_002166.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02000484.hg.1 TC02000041.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02000484.hg.1 ENST00000234091.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02000484.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02000484.hg.1 NM_002166.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011356.hg.1 ENST00000462623.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02000483.hg.1 TC02000041.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02000483.hg.1 ENST00000234091.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02000483.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02000483.hg.1 NM_002166.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02000478.hg.1 TC02000041.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02000478.hg.1 ENST00000234091.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02000478.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02000478.hg.1 NM_002166.4 Refseq
          Affymetrix Human Clariom D Assay TC0300011356.hg.1 ENST00000462623.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145332.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145332.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145332.hg.1 NM_002166.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200145332.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145332.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145332.hg.1 NM_002166.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200145332.hg.1 TC0200006627.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200145340.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145340.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145340.hg.1 NM_002166.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200145340.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145340.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145340.hg.1 NM_002166.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200145340.hg.1 TC0200006627.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200145333.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145333.hg.1 ENST00000331129.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145333.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145333.hg.1 NM_002166.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200145333.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145333.hg.1 ENST00000331129.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145333.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145333.hg.1 NM_002166.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200145333.hg.1 TC0200006627.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200145334.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145334.hg.1 ENST00000331129.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145334.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145334.hg.1 NM_002166.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200145334.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145334.hg.1 ENST00000331129.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145334.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145334.hg.1 NM_002166.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200145334.hg.1 TC0200006627.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200145336.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145336.hg.1 ENST00000331129.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145336.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145336.hg.1 ENST00000472142.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145336.hg.1 NM_002166.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200145336.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145336.hg.1 ENST00000331129.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145336.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145336.hg.1 ENST00000472142.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145336.hg.1 NM_002166.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200145336.hg.1 TC0200006627.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200145339.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145339.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145339.hg.1 ENST00000472142.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145339.hg.1 NM_002166.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200145339.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145339.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145339.hg.1 ENST00000472142.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200145339.hg.1 NM_002166.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200145339.hg.1 TC0200006627.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 ENST00000234091.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 ENST00000331129.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 ENST00000331129.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 ENST00000331129.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 ENST00000331129.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 ENST00000396290.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 ENST00000472142.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 ENST00000472142.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 ENST00000472142.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 NM_002166.4 Refseq
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 NM_002166.4 Refseq
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 NM_002166.4 Refseq
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 NM_002166.4 Refseq
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 NM_002166.4 Refseq
          Affymetrix Human Clariom D Assay TC0200006627.hg.1 NM_002166.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 129260 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline