| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016016.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016016.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016016.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016016.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016016.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016016.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016016.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016016.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016016.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016016.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016016.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016016.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052294.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052294.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052294.hg.1 |
TC01003347.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052292.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052292.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052292.hg.1 |
TC01003347.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052296.hg.1 |
ENST00000255024.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052296.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052296.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052296.hg.1 |
TC01003347.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052298.hg.1 |
ENST00000255024.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052298.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052298.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052298.hg.1 |
TC01003347.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052297.hg.1 |
ENST00000255024.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052297.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052297.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052297.hg.1 |
TC01003347.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052295.hg.1 |
ENST00000255024.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052295.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052295.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052295.hg.1 |
TC01003347.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052299.hg.1 |
ENST00000255024.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052299.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052299.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052299.hg.1 |
TC01003347.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100016016.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016016.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016016.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016016.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016016.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016016.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016016.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016016.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016016.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016016.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016016.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016016.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100221512.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100221512.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100221512.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100221512.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100221512.hg.1 |
TC0100016016.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100221513.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100221513.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100221513.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100221513.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100221513.hg.1 |
TC0100016016.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100221510.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100221510.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100221510.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100221510.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100221510.hg.1 |
TC0100016016.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100221514.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100221514.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100221514.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100221514.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100221514.hg.1 |
TC0100016016.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100221511.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100221511.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100221511.hg.1 |
ENST00000368223.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100221511.hg.1 |
NM_006617.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100221511.hg.1 |
TC0100016016.hg.1 |
Clariom D TC |