Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19020668.hg.1 |
ENST00000248668.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19020668.hg.1 |
NM_020862.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19020668.hg.1 |
TC19001515.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011947.hg.1 |
ENST00000248668.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011947.hg.1 |
ENST00000248668.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011947.hg.1 |
ENST00000248668.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011947.hg.1 |
ENST00000248668.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011947.hg.1 |
NM_020862.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011947.hg.1 |
NM_020862.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011947.hg.1 |
NM_020862.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011947.hg.1 |
ENST00000248668.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011947.hg.1 |
ENST00000248668.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011947.hg.1 |
ENST00000248668.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011947.hg.1 |
ENST00000248668.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011947.hg.1 |
NM_020862.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011947.hg.1 |
NM_020862.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011947.hg.1 |
NM_020862.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900206174.hg.1 |
ENST00000248668.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900206174.hg.1 |
NM_020862.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900206174.hg.1 |
TC1900011947.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900206174.hg.1 |
ENST00000248668.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900206174.hg.1 |
NM_020862.1 |
Refseq |