Search
Search

IFNA1| Gene Symbol: IFNA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024013.3 | NP_076918.1 | 1 | 748 | 183 | |
| GenBank mRNA | AB578886.1 | - | 1 | 746 | 183 | |
| V00537.1 | - | 1 | 735 | 183 | ||
| V00538.1 | - | 1 | 733 | 183 | ||
Set Membership: |
|