| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000462191.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000462191.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000462191.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000470158.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000470158.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000470158.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000470158.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012088.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022783.hg.1 |
ENST00000368891.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022783.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022783.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022783.hg.1 |
TC10001717.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012780.hg.1 |
ENST00000490852.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022785.hg.1 |
ENST00000368891.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022785.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022785.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022785.hg.1 |
TC10001717.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022786.hg.1 |
ENST00000368891.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022786.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022786.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022786.hg.1 |
TC10001717.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022787.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022787.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022787.hg.1 |
TC10001717.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000425266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000425266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000425266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000182710.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000182710.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000182710.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000182710.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000182710.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000182710.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000182710.hg.1 |
TC1000012088.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000182708.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000182708.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000182708.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000182708.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000182708.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000182708.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000182708.hg.1 |
TC1000012088.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000182712.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000182712.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000182712.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000182712.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000182712.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000182712.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000182712.hg.1 |
TC1000012088.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000425266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000425266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000425266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000462191.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000462191.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000462191.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000470158.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000470158.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000470158.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000470158.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012088.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000182713.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000182713.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000182713.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000182713.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000182713.hg.1 |
TC1000012088.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300012780.hg.1 |
ENST00000490852.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000182711.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000182711.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000182711.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000182711.hg.1 |
ENST00000368891.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000182711.hg.1 |
ENST00000481909.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000182711.hg.1 |
NM_024942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000182711.hg.1 |
TC1000012088.hg.1 |
Clariom D TC |