| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02021864.hg.1 |
TC02001393.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02021864.hg.1 |
ENST00000313965.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02021864.hg.1 |
NM_152614.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02021863.hg.1 |
TC02001393.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02021863.hg.1 |
ENST00000313965.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02021863.hg.1 |
NM_152614.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011070.hg.1 |
ENST00000313965.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011070.hg.1 |
ENST00000313965.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011070.hg.1 |
ENST00000313965.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011070.hg.1 |
ENST00000313965.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011070.hg.1 |
NM_152614.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011070.hg.1 |
NM_152614.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011070.hg.1 |
NM_152614.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011070.hg.1 |
NM_152614.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011070.hg.1 |
NM_152614.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200179468.hg.1 |
ENST00000313965.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200179468.hg.1 |
NM_152614.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200179468.hg.1 |
ENST00000313965.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200179468.hg.1 |
NM_152614.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200179468.hg.1 |
TC0200011070.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200011070.hg.1 |
ENST00000313965.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011070.hg.1 |
ENST00000313965.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011070.hg.1 |
ENST00000313965.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011070.hg.1 |
ENST00000313965.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011070.hg.1 |
NM_152614.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011070.hg.1 |
NM_152614.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011070.hg.1 |
NM_152614.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011070.hg.1 |
NM_152614.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011070.hg.1 |
NM_152614.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200179467.hg.1 |
NM_152614.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200179467.hg.1 |
NM_152614.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200179467.hg.1 |
TC0200011070.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200179469.hg.1 |
ENST00000313965.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200179469.hg.1 |
NM_152614.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200179469.hg.1 |
ENST00000313965.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200179469.hg.1 |
NM_152614.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200179469.hg.1 |
TC0200011070.hg.1 |
Clariom D TC |