Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016341.hg.1 |
TC06001422.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016341.hg.1 |
ENST00000333151.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016341.hg.1 |
NM_021066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007376.hg.1 |
ENST00000631369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011225.hg.1 |
ENST00000333151.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011225.hg.1 |
ENST00000333151.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011225.hg.1 |
ENST00000625569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011225.hg.1 |
NM_021066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011225.hg.1 |
NM_021066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011225.hg.1 |
NM_021066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175712.hg.1 |
TC0600011225.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600175711.hg.1 |
ENST00000333151.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175711.hg.1 |
NM_021066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175711.hg.1 |
ENST00000333151.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175711.hg.1 |
NM_021066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175711.hg.1 |
TC0600011225.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600175710.hg.1 |
ENST00000333151.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175710.hg.1 |
NM_021066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175710.hg.1 |
ENST00000333151.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175710.hg.1 |
NM_021066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175710.hg.1 |
TC0600011225.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600011225.hg.1 |
ENST00000333151.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011225.hg.1 |
ENST00000333151.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011225.hg.1 |
ENST00000625569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011225.hg.1 |
NM_021066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011225.hg.1 |
NM_021066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011225.hg.1 |
NM_021066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007376.hg.1 |
ENST00000631369.1 |
Ensembl |