Search
Search

ZNF816| Gene Symbol: ZNF816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031665.2 | NP_001026835.1 | 4 - 5 | 541 | 102 | |
| NM_001202456.1 | NP_001189385.1 | 3 - 4 | 400 | 102 | ||
| NM_001202457.1 | NP_001189386.1 | 3 - 4 | 390 | 102 | ||
| GenBank mRNA | AK291990.1 | - | 3 - 4 | 304 | 102 | |
| BC105739.1 | - | 3 - 4 | 247 | 40026775 | 102 | |
| BC105740.1 | - | 3 - 4 | 247 | 40026776 | 102 | |
| BC105930.1 | - | 3 - 4 | 247 | 40026773 | 102 | |
| DA473052.1 | - | 3 - 4 | 400 | 102 | ||
Set Membership: |
|