Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04192987_gH
          See other SPATA31A1 GE Assays ›
          Gene Symbol:
          SPATA31A1
          Gene Name:
          SPATA31 subfamily A member 1
          Gene Aliases:
          C9orf36, C9orf36A, FAM75A1, FAM75A2, SPATA31A2
          Chromosome Location:
          Chr.9: 39355667 - 39361962 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04192987_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SPATA31A1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001085452.2 NP_001078921.2 3 - 4 371 99
          XM_011518006.1 XP_011516308.1 3 - 4 409 99
          GenBank mRNA AK302569.1 - 3 - 4 371 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900007211.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09000228.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s)

          spermatogenesis
          cell differentiation

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06712763 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16180542 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 765873 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 35813 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 595713 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 387119 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 476388 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 260854 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 205804 AF054180.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 205804 AL832713.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 205804 BC043147.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 205804 NM_203282.2 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CNV Microarray 2x400K 74719 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 357025 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 113799 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 227066 AB037770.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 227066 AK131401.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 227066 BC103943.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 227066 BC103944.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 227066 BC103945.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 227066 BC103946.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 227066 NM_020787.1 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CNV Microarray 2x400K 316182 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 273517 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 40052 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P435874 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 8745 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 31811 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P435874 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3382403 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P435874 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3217364 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P435874 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P435874 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06712763 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16180542 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8155341 - -
          Affymetrix Human Gene 1.0 ST Array 8161345 - -
          Affymetrix Human Gene 1.0 ST Array 8161343 - -
          Affymetrix Human Gene 1.0 ST Array 8161472 - -
          Affymetrix Human Gene 1.0 ST Array 8156255 - -
          Affymetrix Human Gene 1.0 ST Array 8155372 - -
          Affymetrix Human Gene 1.0 ST Array 8161475 - -
          Affymetrix Human Gene 1.0 ST Array 8155374 - -
          Affymetrix Human Gene 1.0 ST Array 8155385 - -
          Affymetrix Human Gene 1.0 ST Array 8155342 - -
          Affymetrix Human Gene 1.0 ST Array 8155343 - -
          Affymetrix Human Gene 1.0 ST Array 8161474 - -
          Affymetrix Human Gene 1.0 ST Array 8155351 - -
          Affymetrix Human Gene 1.0 ST Array 8162215 - -
          Affymetrix Human Gene 1.0 ST Array 8161344 - -
          Affymetrix Human Gene 1.0 ST Array 8161414 - -
          Affymetrix Human Gene 1.0 ST Array 8155344 - -
          Affymetrix Human Gene 1.0 ST Array 8155384 - -
          Affymetrix Human Gene 1.0 ST Array 8161413 - -
          Affymetrix Human Gene 1.0 ST Array 8155375 - -
          Affymetrix Human Gene 1.0 ST Array 8156254 - -
          Affymetrix Human Gene 1.0 ST Array 8155373 - -
          Affymetrix Human Gene 1.0 ST Array 8155353 - -
          Affymetrix Human Gene 1.0 ST Array 8161342 - -
          Affymetrix Human Gene 1.0 ST Array 8155387 - -
          Affymetrix Human Gene 1.0 ST Array 8155352 - -
          Affymetrix Human Gene 1.0 ST Array 8161473 - -
          Affymetrix Human Gene 1.0 ST Array 8155350 - -
          Affymetrix Human Gene 1.0 ST Array 8161411 - -
          Affymetrix Human Gene 1.0 ST Array 8155386 - -
          Affymetrix Human Gene 1.1 ST Array 8161341 - -
          Affymetrix Human Gene 1.1 ST Array 8156253 - -
          Affymetrix Human Gene 1.1 ST Array 8155340 - -
          Affymetrix Human Gene 1.1 ST Array 8161409 - -
          Affymetrix Human Gene 1.1 ST Array 8155383 - -
          Affymetrix Human Gene 1.1 ST Array 8161471 - -
          Affymetrix Human Gene 1.1 ST Array 8155349 - -
          Affymetrix Human Gene 1.1 ST Array 8155371 - -
          Affymetrix Human Transcriptome Array 2.0 PSR09002643.hg.1 ENST00000377647.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09002643.hg.1 NM_001085452.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09002643.hg.1 TC09000228.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09002642.hg.1 ENST00000377647.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09002642.hg.1 NM_001085452.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09002642.hg.1 TC09000228.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0900149120.hg.1 ENST00000377647.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900149120.hg.1 NM_001085452.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900149120.hg.1 ENST00000377647.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900149120.hg.1 NM_001085452.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900149120.hg.1 TC0900007211.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 215935_at - -
          Human Genome U133 Plus 2.0 Array 215935_at - -
          Human Genome U133 Plus 2.0 Array 215935_PM_at - -
          Human Genome U133 Plus 2.0 Array 215935_PM_at - -
          Human Genome U133A 2.0 Array 215935_PM_at - -
          Human Genome U133A 2.0 Array 215935_PM_at - -
          Human Genome U133A 2.0 Array 215935_at - -
          Human Genome U133A 2.0 Array 215935_at - -
          Human Genome U95 Set 34511_PM_s_at - -
          Human Genome U95 Set 34511_PM_s_at - -
          Human Genome U95 Set 34511_s_at - -
          Human Genome U95 Set 34511_s_at - -
          Human Genome U133 Set 215935_at - -
          Human Genome U133 Set 215935_PM_at - -
          Human Genome U133 Set 215935_PM_at - -
          Human Genome U133 Set 215935_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A038595 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A038595 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1709379 NM_001085452.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1709379 NM_001040065.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1709379 AK302569.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1709379 BC146855.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2150446 NM_001085452.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2150446 NM_001040065.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2150446 BC146855.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1766894 - -
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1774527 - -
          HumanHT-12 V4.0 expression beadchip ILMN_2150446 NM_001085452.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2150446 NM_001040065.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2150446 BC146855.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1709379 NM_001085452.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1709379 NM_001040065.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1709379 AK302569.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1709379 BC146855.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2150446 NM_001085452.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2150446 NM_001040065.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2150446 BC146855.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2150446 NM_001085452.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2150446 NM_001040065.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2150446 BC146855.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1774527 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1709379 NM_001085452.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1709379 NM_001040065.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1709379 AK302569.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1709379 BC146855.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1766894 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2150446 NM_001085452.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2150446 NM_001040065.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2150446 BC146855.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1652939 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1766894 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2150446 NM_001085452.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2150446 NM_001040065.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2150446 BC146855.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1652939 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1766894 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2150446 NM_001085452.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2150446 NM_001040065.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2150446 BC146855.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1709379 NM_001085452.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1709379 NM_001040065.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1709379 AK302569.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1709379 BC146855.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1709379 NM_001085452.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1709379 NM_001040065.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1709379 AK302569.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1709379 BC146855.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2150446 NM_001085452.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2150446 NM_001040065.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2150446 BC146855.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline