| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019098.hg.1 |
ENST00000277874.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019098.hg.1 |
ENST00000336152.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019098.hg.1 |
ENST00000371719.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019098.hg.1 |
NM_000872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019098.hg.1 |
NM_019859.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019098.hg.1 |
NM_019860.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019098.hg.1 |
TC10001515.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006904.hg.1 |
ENST00000538670.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006904.hg.1 |
ENST00000543321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006904.hg.1 |
ENST00000543321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006904.hg.1 |
ENST00000543321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006904.hg.1 |
ENST00000543321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006904.hg.1 |
ENST00000543321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006904.hg.1 |
ENST00000543321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006904.hg.1 |
ENST00000543321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006904.hg.1 |
ENST00000624664.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006904.hg.1 |
ENST00000624664.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006904.hg.1 |
ENST00000624664.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006904.hg.1 |
ENST00000624664.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006904.hg.1 |
NR_002774.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006904.hg.1 |
NR_002774.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006904.hg.1 |
NR_002774.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006904.hg.1 |
NR_002774.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019097.hg.1 |
ENST00000277874.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019097.hg.1 |
ENST00000336152.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019097.hg.1 |
ENST00000371719.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019097.hg.1 |
ENST00000371721.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019097.hg.1 |
NM_000872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019097.hg.1 |
NM_019859.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019097.hg.1 |
NM_019860.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019097.hg.1 |
TC10001515.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
ENST00000277874.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
ENST00000277874.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
ENST00000277874.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
ENST00000277874.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
ENST00000277874.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
ENST00000336152.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
ENST00000336152.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
ENST00000336152.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
ENST00000336152.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
ENST00000336152.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
ENST00000336152.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
ENST00000371719.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
ENST00000371719.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
ENST00000371719.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
ENST00000371719.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
ENST00000371719.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
NM_000872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
NM_000872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
NM_000872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
NM_000872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
NM_000872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
NM_000872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
NM_019859.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
NM_019859.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
NM_019859.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
NM_019859.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
NM_019859.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
NM_019859.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
NM_019859.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
NM_019860.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
NM_019860.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
NM_019860.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
NM_019860.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
NM_019860.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011398.hg.1 |
NM_019860.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019099.hg.1 |
NM_000872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019099.hg.1 |
NM_019859.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019099.hg.1 |
NM_019860.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019099.hg.1 |
TC10001515.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1000177431.hg.1 |
NM_000872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177431.hg.1 |
NM_019859.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177431.hg.1 |
NM_019860.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177431.hg.1 |
NM_000872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177431.hg.1 |
NM_019859.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177431.hg.1 |
NM_019860.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177431.hg.1 |
TC1000011398.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200006904.hg.1 |
ENST00000538670.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006904.hg.1 |
ENST00000543321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006904.hg.1 |
ENST00000543321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006904.hg.1 |
ENST00000543321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006904.hg.1 |
ENST00000543321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006904.hg.1 |
ENST00000543321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006904.hg.1 |
ENST00000543321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006904.hg.1 |
ENST00000543321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006904.hg.1 |
ENST00000624664.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006904.hg.1 |
ENST00000624664.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006904.hg.1 |
ENST00000624664.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006904.hg.1 |
ENST00000624664.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006904.hg.1 |
NR_002774.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006904.hg.1 |
NR_002774.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006904.hg.1 |
NR_002774.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006904.hg.1 |
NR_002774.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
ENST00000277874.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
ENST00000277874.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
ENST00000277874.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
ENST00000277874.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
ENST00000277874.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
ENST00000336152.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
ENST00000336152.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
ENST00000336152.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
ENST00000336152.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
ENST00000336152.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
ENST00000336152.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
ENST00000371719.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
ENST00000371719.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
ENST00000371719.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
ENST00000371719.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
ENST00000371719.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
NM_000872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
NM_000872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
NM_000872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
NM_000872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
NM_000872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
NM_000872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
NM_019859.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
NM_019859.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
NM_019859.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
NM_019859.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
NM_019859.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
NM_019859.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
NM_019859.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
NM_019860.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
NM_019860.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
NM_019860.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
NM_019860.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
NM_019860.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011398.hg.1 |
NM_019860.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177430.hg.1 |
ENST00000277874.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177430.hg.1 |
ENST00000336152.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177430.hg.1 |
ENST00000371719.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177430.hg.1 |
NM_000872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177430.hg.1 |
NM_019859.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177430.hg.1 |
NM_019860.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177430.hg.1 |
ENST00000277874.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177430.hg.1 |
ENST00000336152.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177430.hg.1 |
ENST00000371719.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177430.hg.1 |
NM_000872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177430.hg.1 |
NM_019859.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177430.hg.1 |
NM_019860.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177430.hg.1 |
TC1000011398.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000177429.hg.1 |
ENST00000277874.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177429.hg.1 |
ENST00000336152.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177429.hg.1 |
ENST00000371719.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177429.hg.1 |
NM_000872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177429.hg.1 |
NM_019859.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177429.hg.1 |
NM_019860.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177429.hg.1 |
ENST00000277874.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177429.hg.1 |
ENST00000336152.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177429.hg.1 |
ENST00000371719.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177429.hg.1 |
NM_000872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177429.hg.1 |
NM_019859.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177429.hg.1 |
NM_019860.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177429.hg.1 |
TC1000011398.hg.1 |
Clariom D TC |